  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Cold-Formed Steel Shakes Up Building Construction
Cold-Formed Steel Shakes Up Building Construction
Written by Chris Burnett | Published: 09.30.2014
steel building constructionSteel is one of the most widely used and recycled materials. There are thousands of grades of steel with equally as many applications. There are also different technologies for manufacturing steel, one of which offers a major advantage in the construction industry: cold-formed steel (CFS).
Hot-rolled steel and cold-formed steel are both structural steels used in building construction. Hot rolled steel is made by rolling steel into thin strips or plates. Hot rolling occurs at very high temperatures, above the recrystallization temperature of the material, producing malleable steel that can be formed in a variety of shapes.
Cold-formed steel, also called light gauge steel is made from steel plate, sheet or strip material. Cold-formed steel can be made by roll forming, the method used for mass production, by stamping, commonly used in the automotive industry, or by press braking. Cold rolling is a metal forming process in which a sheet of metal is pressed through a pair of rolls to reduce thickness, increase strength and improve surface finish. The cold rolling process happens at ambient temperature, below the steel’s recrystallization temperature. This process changes the mechanical properties of the steel to produce uniform, lightweight steel with a high strength-to-weight ratio. Cold-formed steel is also easy to fabricate, transport, and install; it is moisture-, fire-, wear- and corrosion-resistant; and it is energy efficient. Read the American Iron and Steel Institute’s Cold-Formed Steel Profile for an in-depth look at the reasons why cold-formed steel is becoming the material of choice in construction.
The Studies
Study results presented at Quake Summit 2014 in Anchorage, Alaska, part of the 10th U.S. National Conference on Earthquake Engineering, provide another reason why cold-formed steel is an advantageous building material—it appears to be nearly earthquake-proof.
CFS-NEES is a three-year research project funded by the George E. Brown Jr. Network for Earthquake Engineering Simulation (NEES) and the steel industry to test the ability of buildings made of lightweight cold-formed steel to withstand an earthquake. The research team is composed of scientists from six universities and design consultants from the steel industry, and is lead by researchers from Johns Hopkins University. The researchers constructed a two-story building using cold-formed steel for its primary beams and columns upon a moving platform known as a “shake table” and subjected it to a series of lab-generated high-magnitude earthquakes. The results were surprising—the structure suffered almost no damage. The team hopes to make the data available to building engineers to help them improve the seismic safety of their designs.
These findings will likely increase the demand for cold-formed steel, and steel producers need to ensure their product meets building material specifications. To achieve high quality steel strip, thickness gauges are used to detect and correct deviations in thickness in real-time. X-ray gauges are the optimal thickness gauge technology to provide the speed and accuracy required to operate a cold rolling mill. X-ray based sensor systems provide extremely accurate high speed/low noise measurements, permitting producers to realize material savings and quality improvements.
Read this Optimization of a Cold Rolling Mill with a High Speed X-ray Thickness Gauge paper to learn about other advantages of an x-ray-based thickness sensor.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
215
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Max Mathews says
    September 27, 2016 at 7:58 am
    Thanks for giving this detailed description in simple words…you can also see different manufacture analysis in Cold-Formed Market @ http://bit.ly/2d52oRR
    Reply
  2. Frank D. Walker says
    September 30, 2016 at 1:28 pm
    Nice article and very informative. Thanks for sharing. For other info on steel building construction, visit us : http://www.steelcobuildings.com/
    Reply
  3. IUP Jindal says
    December 21, 2019 at 9:16 am
    Thanks for sharing the blog!!
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Testing, Testing, 1-2-3 : Numismatic Testing Methods
Technology Focus: X-Ray Photoelectron Spectroscopy (XPS)
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook