  • Contact
Accelerating ScienceAnalyzing Metals / Precious Metals / Could Platinum Group Metals Save Your Life?
Could Platinum Group Metals Save Your Life?
Written by Christopher Shaffer | Published: 02.10.2015
Editor’s Note:  This post is Part 5 of a series that examines the wide range of industrial, medical, and electronic applications of the six platinum group metals (PGMs), which are platinum (Pt), palladium (Pd), rhodium (Rh), iridium (Ir), ruthenium (Ru) and osmium (Os).
Most pacemakers contain at least two platinum-iridium electrodes.
Most pacemakers contain at least two platinum-iridium electrodes.
The most famous Platinum Group Metals (PGMs), platinum and palladium, are best known as jewelry and as primary components in automotive catalytic converters, but they’re also biocompatible materials that have several less-known but very important, even life-saving, medical applications. Here’s a summary; for more details, please visit the the International Platinum Group Metals Association web site.
Cancer
Platinum and palladium have been active chemotherapy agents since the 1970s and an estimated half or more of all cancer patients who receive chemotherapy are treated with drugs containing platinum. Cisplatin, carboplatin and oxaliplatin are all platinum compounds that have been particularly successful in the treatment of testicular and ovarian cancers.
Platinum and palladium also play active roles in cancer radiation therapy. Palladium-103, a radioactive isotope of palladium, shows promise for the treatment of breast and prostate cancer. The outpatient procedure involves permanently implanting small time-release seeds of palladium-103 directly into the tumor to deliver the highest possible dose directly to the source while sparing the surrounding tissue.
Medical devices and implants
Platinum is inert, rarely causes allergic reactions, has good electrical conductivity, and can be fabricated into tiny components, all qualities that make it an ideal material for pacemakers, stents, implantable defibrillators, and catheters.  Most pacemakers contain at least two platinum-iridium electrodes. Platinum marker bands and guide wires are commonly used to place stents, and now even the stents themselves may be made with platinum alloys. Platinum marker bands are also placed on catheters and guide wires to help surgeons with device placement. Platinum-iridium electrodes are used in neuromodulators such as aural and retinal implants as well as devices used to treat Parkinson’s disease and chronic pain.
Dental
Palladium, and occasionally platinum, ruthenium or iridium are used to enhance the strength and durability of dental inlays, crowns and bridges. The PGMs are usually mixed with gold, silver, copper or zinc to produce alloys for dentistry.
Testing Technologies
Many electronic and automotive devices containing PGMs are recycled to recover and reuse the metal, but some of the medical devices mentioned above, such as defibrillators, are recyclable as well. Whether from primary or secondary sources, PGMs must be meticulously refined to separate the PGMs from base metals and contaminants, analyzed for purity, and fabricated to produce medical-grade material. Lab-based PGM analysis includes careful sample preparation and multi-step assaying to detect all minor and trace elements and to ensure the PGM is of pure enough quality for medical applications. Testing technologies to determine precious metal concentrations may include:
ICP-OES: Inductively Coupled Plasma – Optical Emission Spectrometry (ICP-OES) , also known as ICP-AES, is an analytical technique used to detect minor and trace elements in a variety of sample types.
ICP-MS ICP-MS is a technique for the analysis and quantification of trace elements in both solid and liquid samples.
X-ray fluorescence (XRF): XRF a non-destructive analytical technique used to determine the qualitative and quantitative elemental composition of materials.
Learn more about each of these elemental analysis techniques.
Share this article
4
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
U.S. Department of Energy Transforms Gold Alloys with Air
Getting to the Heart of Surface Analysis
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook