  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / Cutting Technology for Knife Blade Metals
Cutting Technology for Knife Blade Metals
Written by Brian Wilson | Published: 05.26.2020
Cutting Technology for Knife Blade MetalsI recently saw an article published by the World Steel Organization about a master Japanese blacksmith who said: “The quality of a Japanese knife lies in its hardness and strength. Ideally, it’s unbreakable, unbendable, and unchippable, but these qualities conflict with one another.”
The article talks about how the secret to knife success involves the master-quality craftsmanship and reverence for perfection in even the smallest of details, including the microstructure of the metal itself.  It was explained that different metals, including steel, have different properties and uses, and craftsmen apply their expertise to bring out the best in their chosen source material.
For blade crafting, one Japanese knife company uses locally-sourced high-carbon steel that has been specifically formulated for making sharp blades. The blacksmiths there believe the ability of steel to adapt to multiple use cases and its ability to be optimized for the qualities needed, such as rust-resistance, is fascinating. He also believes that there are scientific reasons why a super sharp Japanese blade makes a difference when chefs prepare foods, especially sushi.
So how can that combination of hardness, strength, and rust-resistance needed for the perfect Japanese knife be attained?  It’s all about the alloying – the combining of two or more metallic elements to enhance metal properties – and working the metal properly.
We previously wrote about the difference between metal strength and stiffness. A material can have high strength and low stiffness. If a metal cracks easily, it has low strength, but if it has low stiffness, it can deflect a high load.  All steel has approximately the same stiffness, but comes in many different strengths depending on the alloying metals used.  Stainless steel comes in more than 100 grades which are created by adding alloys such as chromium, silicon, nickel, carbon, nitrogen, and manganese to impart properties such as heat resistance, strength, flexibility, and ductility.
Also, a metal’s properties such as weldability and heat or corrosion resistance depend on the carbon content in steel. Carbon works as a hardening agent. Increasing carbon content increases hardness and strength, but increases brittleness as well.  (You can learn more about the metallurgy of steel and its carbon content on thefabricator.com.)
Of course one must get the alloy recipe right, mixing together the precise amount of each element.  If not, the quality will suffer, and the product can fail. A steel blade that breaks during use can possibly hurt the user if it snaps off and cuts someone.  And a blade that rusts would certainly tarnish a manufacturer’s reputation.
I’m kind of a knife nerd, or I probably just own too many. Looking in my collection there’s a representation of the most common knife steels, and I’ve noted the typical C% for reference:
  •        440C     ‒     0.95-1.20%
  •        1095     ‒      0.95%
  •        154CM ‒      1.05%
  •        420        ‒     0.15% min
  •        N690     ‒     1.0%
  •        AUS-8   ‒     0.75%
To help ensure quality and product integrity, XRF handheld analyzers are used to confirm the elements in the metal. X-ray fluorescence (XRF) is a proven technology for the elemental analysis of specialty alloys to ensure the correct alloys are combined in the right percentages and the finished material meets precise manufacturing specifications. This is critical for quality control and quality assurance (QA/QC) of incoming materials and outgoing finished goods.  (Read about how X-ray fluorescence (XRF) technology is increasingly being adopted to identify unknown materials and verify material composition throughout the automotive product development and manufacturing process.)
There is one caveat though. An XRF analyzer with light element feature is a great tool for measuring all alloying elements with exception of carbon. But carbon is what drives hardenability and ultimately edge retention and ease of sharpening so for exacting chemistry or verifying a knife blade prior to shaping, LIBs would be the best option.
Laser Induced Breakdown Spectroscopy (LIBS) is the analytical technique using a high-focused laser to determine the chemical composition of materials. The technique is available in a portable handheld analyzer and is capable of measuring elements, including carbon, in the field for material identification. However, since the LIBS technique utilizes a high-focused laser that interacts with the surface of a material, it forms a plasma in which the material is broken down into single elements. A laser pulse is produced by the analyzer and pointed at the sample surface. The surface is ablated and enters the plasma.  Most, if not all blade manufacturers, would want to avoid the sample prep and arc strike on a finished blade. So if carbon is important to the finished product, LIBS should be used prior to forming the blade itself.
In the article, the blacksmith said that it is important to constantly make adjustments during the forging process of the knife.  “Knowing how to bring these qualities out of the steel and adjust them at a fine level is something that can only be gained through first-hand experience.”
But before getting to the forging stage, it’s important that they are working with the right materials.  And according to the article, the quality of your sushi could depend upon it.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
198
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Brian Wilson
Business Development Manager, Energy Markets
Field and Safety Instruments
Thermo Fisher Scientific
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Alloy 617 Makes the Grade
Is Platinum a Good Choice for Jewelry?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook