  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / How to Get Accurate Silicon Readings when Analyzing Silicon in Steel
How to Get Accurate Silicon Readings when Analyzing Silicon in Steel
Written by Mathieu Bauer | Published: 08.11.2021
How to Get Accurate Silicon Readings when Analyzing Silicon in Steel Typical metal alloys that are at risk of sulfidation corrosion are carbon steels. At high temperature, these alloys will corrode leading to metal wall thinning when exposed to sulfur compounds present in crude oil and can cause failures. Those failures can result in leaks, early replacements, unplanned outages, and incidents potentially resulting in loss of property and injury to workers.
To help prevent these incidents from occurring, silicon analysis of carbon steel is done. Handheld XRF instruments are used in as Positive Material Identification (PMI) programs to analyze carbon steel  and verify elements where sulfidic corrosion, is a concern.  X-ray Fluorescence (XRF) technology is a non-destructive analytical technique used to determine the elemental composition of materials.
Data quality objectives dictate the sample preparation requirements and the minimum analysis time used when analyzing the steel. Surface oxidation can affect the accuracy of the reading when performing an XRF analysis. As the chromium concentration in the metal gets lower, the oxidation will get worse. It is imperative to remove any corrosion in order to ensure an accurate reading.
In addition to oxidation, there can often be paint or oil or grease on the surface. Paint typically contains metals, such as titanium, zinc or calcium, that can impact the results of analysis. Grease can contain molybdenum and other additives.
In order to get the accurate silicon readings, all surface contamination must be removed in the area to be analyzed.
The fastest way to prepare an oxidized surface is to use a right angle die grinder equipped with the proper abrasive media, such as a zirconia/alumina grinding disc.
Since data quality objectives dictate the sample preparation requirements and the minimum analysis time used, we analyzed samples against certified reference standards after ensuring the surface was clean and clear of any contaminants that could introduce silicon or other elements to our analysis.
Depending on the concentration of silicon and the desired precision, the samples were analyzed for 9 to 33 seconds using both the main filter (3 seconds) and light filter (between 6 and 30 seconds) after thorough surface preparation. We were able to identify both pipe material that passed inspection as it was not susceptible to sulfidic corrosion, and pipe material that failed and would be susceptible to sulfidic corrosion because it did not reach the recommended threshold of 0.1% Si in carbon steel.
For additional details, including methods and instruments used, correlation curves, measurement repeatability data, results and conclusion, read the application note: Analysis of Silicon in Steel to Prevent Sulfidic Corrosion Using the Niton XL5 Plus XRF Analyzer
Resources:
  • Application Note: Analysis of Silicon in Steel to Prevent Sulfidic Corrosion Using the Niton XL5 Plus XRF Analyzer
  • Application Note: Conducting Retroactive PMI
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
An Ideal Galvanized Steel Sample Collection Process
How to Improve Your Scrap Metal Recycling Business with Handheld Analyzers
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook