  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / INFOGRAPHIC: Austenitic Stainless Steels
INFOGRAPHIC: Austenitic Stainless Steels
Written by Marlene Gasdia-Cochrane | Published: 12.15.2015
Editor’s Note:  This is Part 1 in a 3-Part Infographic Series on Stainless Steels.
Austenitic Steel infographicDid you know there are more than 100 grades of stainless steel? Stainless steel is made by adding varying amounts of chromium (Cr) and other alloying elements such as nickel (Ni) to iron and carbon to create a corrosion-resistant product. We will cover three of the five primary grades of stainless steel (austenitic,ferritic, martensitic, duplex, and precipitation hardening) in infographic form. This week we will address Austenitic Steel.
Austenitic stainless steels, named after the UK metallurgist Sir W. C. Roberts-Austen, contain high amounts of chromium and nickel and are the most corrosion resistant, ductile, and weldable type of stainless steel. Take a look at this infographic and get a snapshot of Austenitic Stainless Steels.
Want to know more about Stainless Steel?
Read Is Stainless Steel Really Stainless?
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
4
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
VIDEO: See How Sinter Feed Gets Blended and Analyzed
INFOGRAPHIC: Ferritic Stainless Steels
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook