  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / INFOGRAPHIC: Bismuth - The Heaviest Among Heavy Metals
INFOGRAPHIC: Bismuth – The Heaviest Among Heavy Metals
Written by Chris Calam | Published: 03.01.2016
Bismuth InfographicLead, a highly toxic heavy metal, is known for its detrimental effects on health, and there are mandates to remove it in many places. In fact, according to the U.S. Geological Survey, “The Safe Drinking Water Act Amendment of 1996, which required that all new and repaired fixtures and pipes for potable water supply be lead free after August 1998, opened a wider market for bismuth as a metallurgical additive to lead-free pipe fittings, fixtures, and water meters.
Elemental bismuth occurs as metallic crystals associated with nickel, cobalt, silver, tin, and uranium sulphide ores.  Number 83 on the periodic table, it is mainly a byproduct of lead ore processing; yet among the heavy metals, it is the heaviest and the only non-toxic.
As a result, bismuth is becoming an ingredient in many metal alloys and products. In fact, researchers in China developed a low-melting-point bismuth-indium-tin-zinc alloy that allows the liquid alloy to be squeezed through tubes that could be used in the 3D printing of electronic components. The new alloy’s low melting point requires little cooling, allowing for faster printing than is currently possible with higher melting point metals currently in use.”
Take a look at this Bismuth infographic and get nine fast facts about this increasingly important metal.
Share this article
59
SHARES
FacebookLinkedinTwitterMail
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Small Spot Analysis, Mapping, and Standardless Analysis Explained
What’s Your Copper, Nickel, Iron, or Gold Worth?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook