  • Contact
Accelerating ScienceAnalyzing Metals / Precious Metals / Jewelry Maker Switches to All Recycled Gold and Silver
Jewelry Maker Switches to All Recycled Gold and Silver
Written by Chris Calam | Published: 07.14.2020
old silver scrap
stock photo of old silver scrap
Pandora recently announced it will stop using newly mined silver and gold in all its jewelry by 2025 and only buy from recycled sources, a move that will greatly reduce carbon emissions for both metals.
According to a statement on the company’s web site, 71% of the silver and gold in Pandora’s jewelry already comes from recycled sources. Shifting completely to recycled silver and gold will reduce CO2 emissions, water usage and other environmental impacts associated with mining new metals. The carbon emissions from sourcing recycled silver are a third of mined silver, while recycling gold emits approximately 600 times less carbon than mining new gold, according to life cycle assessments.
Silver is the most used material in Pandora jewelry, but the company also uses smaller volumes of gold, palladium, copper and man-made stones.
“Silver and gold are beautiful jewelry materials that can be recycled forever without losing their quality. Metals mined centuries ago are just as good as new. They will never tarnish or decay. We wish to help develop a more responsible way of crafting affordable luxury like our jewelry, and prevent that these fine metals end up in landfills. We want to do our part to build a more circular economy,” said CEO Alexander Lacik.
Pandora says it will work with its suppliers to guarantee a sufficient supply of responsibly sourced recycled silver, certified according to leading supply chain initiative standards such as the Responsible Jewelry Council. Pandora will also engage with key stakeholders in the supply chain to explore opportunities for increasing the availability of recycled silver and improve production standards.
Today, around 15% of the world’s silver supply comes from recycled sources. More than half of the recycled silver comes from industry, where the metal is used in chemical production, electronics and for other purposes.
Recycling efforts must be combined with careful elemental analysis of the recovered metal to determine its exact chemical composition and to ensure the metal is free from contaminants or hazardous materials. X-ray fluorescence (XRF), a non-destructive analytical technique used to determine qualitative and quantitative analysis of materials, including precious metals, is a widely-used technology for this type of analysis. XRF analyzers determine the chemistry of a sample by measuring the fluorescent (or secondary) x-ray emitted from a sample when it is excited by a primary x-ray source. Because this fluorescence is unique to the elemental composition of the sample, XRF is an excellent technology for qualitative and quantitative analysis of the material composition.
Precious metal analyzers are especially useful in providing a fast, accurate, and most importantly, nondestructive method to test the purity and chemistry of all precious metals, including silver and gold.
Share this article
FacebookLinkedinTwitterMail
gold metallic number 5
5 Precious Metals Analysis Methods
We’ve previously written about the four primary precious m... by Juan Pablo Zuluaga / 05.15.2025
Read More
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane, Editor / 11.26.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
How to Choose the Right Elemental Analyzer: XRF vs LIBS
Reader Asks About Measuring Coatings on Steel
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook