HF Zinc-Aluminum Alloy is Unique
The HF Zinc-Aluminum Alloy is based on ZAMAK alloys, one of two groups of zinc casting alloys made of approximately 4% aluminum that provide excellent die castability. The other family is the ZA alloys, high-strength alloys which contain higher amounts of aluminum.
The HF Zinc-Aluminum Alloy was developed through the joint efforts of the Cast Metals Coalition, the North American Die Casting Association (NADCA), the International Zinc Association (IZA), and the U.S. Department of Energy. According to the IZA, the HF Zinc-Aluminum Alloy has 40% more fluidity than Alloy 7, which until now was one of the most successful high fluidity zinc alloys. The new HF Alloy also has comparable physical, mechanical, and corrosion properties to Alloy 7 and Alloy 3, comparable fluidity and castability to Superloy/GDSL, and is best suited for casting parts with section thickness less than 0.45 mm. It is also designed for parts that are difficult to fill or that have high surface finish requirements. The new alloy exceeds minimum thickness targets and allows for casting parts with a wall thickness as thin as 0.25 mm.
Die castings are an important product of the metalworking industry, found in everything from cars to electronics to building materials. Die castings made from zinc alloys offer significant advantages over those made from polymers or other metals, including:
- Strength, durability, formability
- Excellent thermal and electrical conductivity
- Castability at moderate temperatures which reduces energy costs and extends tool life
- Work with almost all casting and finishing processes
- Easier and minimized machining
- High casting fluidity
- Recyclable
Each of the different zinc casting alloys requires a very specific formulation to produce the expected characteristics. HF Zinc-Aluminum Alloy, for example, also contains magnesium, copper, iron, lead, cadmium, and tin. The exact percentages of each element, as well as the purity of the zinc and the absence of contaminants must be verified throughout the manufacturing process to guarantee the integrity of the finished product.
Optical Emission Spectrometry (OES) is a widely-used, industry-established technology for accomplishing the process and quality control required for metallurgy. OES instruments are capable of providing rapid qualitative and quantitative analysis to correct chemistry inconsistent in real-time and are an optimal solution for zinc casting alloy elemental analysis.
Hi,
I tried to find manufacturers / suppliers of Superloy. But not found. Umicore does not produce this alloy. Do you have any idea where I can get Superloy?
Best regards
M. Wegebauer
Thank you for your inquiry, M. Wegabauer. Our author recommends that you have a look at the website “TotalMateria” – a comprehensive material database for metals. It contains a module called “Suppliers” that may be helpful.
Your best bet is to go to this website page:
http://www.totalmateria.com/page.aspx?ID=Components&LN=EN
and click “Try for Free
Best regards.