  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / New Reduced Platinum Catalyst for Catalytic Converters
New Reduced Platinum Catalyst for Catalytic Converters
Written by Mark Lessard | Published: 06.12.2018
Catalytic converters are pollution control devices coated with chemicals and a combination of the platinum group metals (PGM) platinum (Pt), rhodium (Rh) and/or palladium (Pd). The PGMs are responsible for the conversion reactions that turn pollutants into harmless gases. Most present-day vehicles that run on gasoline, including more than 98% of new cars sold worldwide each year, as well as  trucks, buses, trains, motorcycles, and planes have exhaust systems with a catalytic converter.
Given the high level of demand and the scarcity of supply, efforts to recover PGMs from spent catalytic converters and other industrial products have been underway for several years. Recycling efforts must be combined with careful elemental analysis of the recovered metal to determine its exact chemical composition and to ensure the metal is free from contaminants or hazardous materials. X-ray fluorescence (XRF), a non-destructive analytical technique used to determine qualitative and quantitative analysis of materials, is a widely-used technology for this type of analysis. 
An alternative to recycling expensive PGMs from catalytic converters is to use less of the metals. Researchers from Washington State University and Tufts University have made progress on this front by demonstrating that a single metal atom can act as a catalyst in converting carbon monoxide into carbon dioxide, the chemical reaction performed by catalytic converters to remove harmful gases from car exhaust.
According to a press release on the Washington State University web site, car companies have struggled to meet strict emissions standards because as engines have become more efficient, their combustion temperature has become lower, making it harder for catalytic converters to work and creating even more harmful emissions. While studying low-temperature catalysts, the researchers tested the ability of single metal atoms to act as catalysts at lower temperatures and demonstrated that the reaction can work with single platinum atoms on a copper oxide support near room temperature. The single platinum atom holds the carbon monoxide in place while the copper oxide supplies the oxygen to convert it into carbon dioxide. Other cheaper metals such as copper will be studied next. The research was published in the journal Nature Catalysis.
Our recent blog post, High-performance Fuel Cells May be Possible by Using Less Platinum, reported on another effort to reduce platinum consumption. As reported on ScienceDaily.com, Johns Hopkins researchers developed a technique to apply a one nanometer thick coating of platinum to a core of much cheaper cobalt, potentially creating a crucial catalyst in new fuel cells that use far less platinum.
The article explains that earlier attempts to plate precious metals on non-precious materials failed due to oxidation of the non-precious metal. Johns Hopkins team successfully suppressed such reactions by introducing carbon monoxide, a gas molecule that strongly binds to cobalt, protecting it from oxidation. Not only did the cobalt-platinum nanoparticles use less platinum; they performed almost 10 times better than platinum alone. The study was published in Nano Letters.
Learn about Portable Metal Manufacturing, Inspection & Recycling Solutions for platinum and other metals, as well as XRF Technology.
Share this article
311
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Portable XRF Analyzers Are a Nondestructive PMI Test Method for Oil and Gas Pipelines
Metallic Glass Can Be Created Using 3D Printing
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook