  • Contact
Accelerating ScienceAnalyzing Metals / Positive Material Identification / Oil Well Construction: Casing and Tubing
Oil Well Construction: Casing and Tubing
Written by Chris Burnett | Published: 03.31.2015
oil well drillingCasing is a series of steel pipes that are run into a drilled oil well to stabilize the well, keep contaminants and water out of the oil stream, and prevent oil from leaching into the groundwater. Casing is installed in layers, in sections of decreasing diameter that are joined together to form casing strings. The five types of casing string are conductor casing, surface casing, intermediate casing, casing liner, and production casing.
Casing is typically made from carbon steel, but as the primary structural component of the well the grade of steel used to make the casing, and the specifications of the finished material, are very important.
Most countries follow American Petroleum Institute (API) standards for the design, manufacturing, testing, and transportation of oil and gas casing used in wells that will be hydraulically fractured. According to API’s Hydraulic Fracturing Operations—Well Construction and Integrity Guidelines, casing must meet strict requirements for compression, tension, collapse, and burst resistance, quality, and consistency. Well casing should withstand hydraulic fracturing pressure, production pressures, and corrosive conditions. Used or reconditioned casing should meet the same API performance requirements as new casing.
Once the casing is in place, tubing is run into the casing to transport the oil or gas. Both casing and tubing are manufactured to certain diameters and wall thicknesses. The Society of Petroleum Engineer’s PetroWIKI site describes the tubing manufacturing process:
API specifies seamless or electric-weld processes for tubing manufacture. Seamless pipe is defined as a wrought steel tubular product made without a welded seam. It is manufactured by hot-working steel or, if necessary, by subsequently cold-finishing the hot-worked product to produce the desired shape, dimensions, and properties. Because of the nature of the manufacturing, the cross section of the tubing wall area may be slightly eccentric and the tubing slightly oval and not perfectly straight.
Electric-welded pipe has one longitudinal seam formed by electric-resistance or electric-induction welding without the addition of filler metal. The edges to be welded are pressed together mechanically, and the heat for welding is generated by the resistance to flow of electric current. The weld seam of electric welded pipe is heat-treated after welding to a minimum temperature of 1,000°F or processed so that no untempered martensite remains.
The spiral and longitudinal welded tubes used in transportation of petroleum are produced from hot working and are made by rolling steel into thin strips or plates. Hot rolling occurs at very high temperatures, above the recrystallization temperature of the material, producing malleable steel that can be formed in a variety of shapes. Cold-formed steel is made by roll forming, a metal forming process in which a sheet of metal is pressed through a pair of rolls to reduce thickness, increase strength and improve surface finish.
Online metal thickness gauges for hot- and cold-rolling mills provide precise, real-time measurements to ensure the finished products meet the exact specifications for the environment in which they function. X-ray gauges are the optimal thickness gauge technology to provide the speed and accuracy required to operate a hot or cold rolling mill. X-ray based sensor systems provide extremely accurate high speed/low noise measurements, permitting producers to realize material savings and quality improvements.
See non-contact thickness gauges for hot- and cold-rolling mills in action- watch the video.
Share this article
156
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Trayson Evans says
    January 27, 2017 at 5:30 pm
    I learned recently that the pipes don’t always go straight. They can be curved to reach different parts of the oil pocket. I would think that this would make it hard to insert and remove the casing and pipes. I think the pipe would be easier because it could just slide along inside the casing.
    Reply
  2. tubing and casing pup joints says
    February 16, 2021 at 11:51 am
    Hi there
    Interesting article. Thanks for sharing with us the knowledge of oil well construction. from this article, I gain more knowledge about casing and tubing. very well defining. keep sharing. well done.
    DIC OIL TOOLS
    https://dic-oiltools.com/tubing-casing.php
    Reply
  3. Nittin Nath Mittal says
    September 15, 2021 at 9:16 am
    Hello
    Excellent article. i want to thankyou for this informative read, i really appreciate sharing this great post. keep up your work.keep sharing.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Mercury Testing and Analysis in the Scrap Metal Industry
Infographic: Aluminium – The Metal Extraordinaire
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook