  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Online Measurement of Oil Coatings Improves Steel Component Manufacturing
Online Measurement of Oil Coatings Improves Steel Component Manufacturing
Written by Chris Burnett | Published: 08.04.2016
sheet metal coilsLubrication oils are commonly used to ensure that the surface of sheet steel behaves consistently during stamping and punching operations. Without a thin layer of lubricant on the steel, the steel may tear, heat up too much, or cause the punch press to jam. However, to prevent unplanned line stoppages, lubricants are often applied in excess, reducing profitability, creating slip hazards in the coil storage areas and generating an additional waste stream to manage. Each of these scenarios can lead to production delays. Online measurement of oil coatings helps metals component manufacturers eliminate costly delays in their process, save raw materials, reduce re-work, and improve product quality.
Traditional oil measurement methods require cutting samples, punching out coupons of exact diameters, and precisely weighing the sample before and after stripping the oil from the surface. While generally accepted, this process reduces yield by taking from the finished coil and happens after the components have been made, too late to make any changes.
An online infrared (IR) light sensor is ideal for measuring hydrocarbon-based lubricants and offers a better solution to ensure that the critical oil layer is thick enough to guarantee consistent production, but not excessively thick to cause waste or pooling in the dies. By positioning an IR light source and detector on the same side of the coated product, a system of optics can be used to measure the intensity of a specific wavelength of reflected light relative to a reference wavelength. By comparing the ratio of the two intensities, a relative measurement of the coating thickness can be made. In general terms, the thicker the coating, the more absorption will occur at the measured wavelength in relation to the reference wavelength and the larger the observed ratio.
A sensor that measures a wide spectrum of IR light is the most appropriate for those production situations that involve multiple coating/substrate compositions. When configured to measure wavelengths associated with an entire range of hydrocarbon bonds, an IR sensor can accurately measure coating weights for nearly every type of coating. Additionally, by using optical light, the sensor is immune to background interference due to air temperature changes between sensor and strip.
By synchronizing source lamp emissions with detector sampling, the sensor will eliminate background light variations and sheet flutter.
The spectrum-based detector, as compared to a two-wavelength IR approach, can measure the coating without the influence of naturally occurring localized variations in zinc coatings, for example. As the light intensity varies across the spangle, the two-wavelength IR sensor would falsely interpret that as a coating change, whereas the spectrum-based detector monitors the influence on the entire spectrum in one single measurement location and is thereby immune to any variation.
The infrared spectrum technology described here addresses the typical influences present in a rolling mill and provides reliable measurements for oil coating thickness in real time. This method results in significant benefits associated with improved coating uniformity, reduced re-work, and elimination of delays while destructive tests are made by sheet suppliers.
(Content from this post originally appeared in the February, 2016 issue of Steel Times International)
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Roy Nuss says
    June 29, 2020 at 12:05 pm
    I looking to measure ATBC (acetyl tributyl citrate) on steel in the 0.5 mg/ft range. Is your IR method able to do that?
    What size area does you method cover?
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      July 27, 2020 at 12:55 pm
      Thanks for your question, Roy. We have answered it here: https://www.thermofisher.com/blog/metals/reader-asks-about-measuring-coatings-on-steel/
      Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
XRF Remains an Important Tool to Combat Steel Pipe Corrosion
Can You Take Portable XRF Analyzers on an Airplane?
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook