  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Platinum Group Metals at Work: Industrial Applications
Platinum Group Metals at Work: Industrial Applications
Written by Christopher Shaffer | Published: 01.27.2015
Editor’s Note:  This is Part 3 of a 6-Part Series on PGMs that runs on Tuesdays.
Fertilizer
Platinum-rhodium catalysts are used in the manufacture of nitric acid, a key ingredient in fertilizer.
The Platinum Group Metals (PGMs) have several unique and valuable properties that make them indispensable for many industrial applications. The PGMs are extremely rare and are considered strategic metals or critical metals because they are necessary for many industrial and technological processes but are in short supply and have no adequate alternatives.
An estimated one-fifth to one fourth of everything we use either contains or is manufactured with platinum, the most valuable of the PGMs. Here’s a summary of just a few of the myriad industrial applications of PGMs; to learn more, visit the International Platinum Group Metals Association web site.
Chemical processing
Thanks to their high temperature stability, melting points and corrosion resistance, platinum, rhodium, iridium and their alloys are used to make industrial crucibles, which are used in virtually every industrial process. High purity conditions are required for these processes and the PGMs, with their high temperature stability, do not contaminate the products being fabricated. Iridium is the preferred material for crucibles used in the production of high purity single crystals of various metal oxides, which have applications in laser technology as well as medical, petroleum, and security industries.
Nitric acid
Platinum-rhodium catalysts are used in the manufacture of nitric acid, a key ingredient in fertilizer. Non-fertilizer applications include the production of explosive-grade ammonium nitrate, adipic acid for making nylon, and toluene diisocyanate for manufacturing polyurethane.
Iridium-ruthenium catalysts are part of the process that makes acetic acid, which has a very wide range of applications including cleaning, photography, fumigation, packaging, paints, adhesives and artificial fibers.
Platinum is also used in the production of paraxylene (PX), and a palladium catalyst is used in the production of purified terephthalic acid (PTA). Both PX and PTA are used to make polyethylene terephthalate (PET) an ingredient in plastics and polyester textiles.
Other applications
  • Ruthenium and iridium are used in the process that produces caustic soda, an ingredient in organic and inorganic chemicals, soaps, and detergents.
  • Palladium is used in the manufacture of the raw materials to make nylon and synthetic rubber thanks to its hydrogen-absorbing qualities.
  • Platinum catalysts are used in the production of silicones, high-octane gas, and petrochemical feedstocks, which are used to make plastics, synthetic rubber, and polyester fibers.
  • Platinum and palladium are widely used in applications requiring stability at high temperatures such as jet engine and missile parts.
  • Platinum and rhodium can be found in thermocouple temperature sensors which are used in the glass, steel, and semiconductor industries. Other platinum wire products may contain iridium, ruthenium and rhenium.
PGMs can be recovered from spent catalysts and other industrial products. Recycling PGMs from industry is much more successful than recovery from consumer electronics. Recycling efforts must be combined with careful elemental analysis of the recovered metal to determine its exact chemical composition and to ensure the metal is free from contaminants or hazardous materials. X-ray fluorescence (XRF) is a widely-used technology for this type of analysis. XRF a non-destructive analytical technique used to determine the elemental composition of materials. XRF analyzers determine the chemistry of a sample by measuring the fluorescent (or secondary) x-ray emitted from a sample when it is excited by a primary x-ray source. Because this fluorescence is unique to the elemental composition of the sample, XRF is an excellent technology for qualitative and quantitative analysis of the material composition.
Learn more about some of the XRF instruments available to improve industrial processes.
Share this article
6
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Video: The Impact of Metals in the Recycling Industry
INFOGRAPHIC: The Metal Titanium
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook