  • Contact
Accelerating ScienceAnalyzing Metals / Positive Material Identification / PMI Testing of 347 Stainless Steel: As-Delivered vs. As-Welded
PMI Testing of 347 Stainless Steel: As-Delivered vs. As-Welded
Written by Brian Wilson | Published: 04.14.2015
In our last article, we discussed how industrial safety regulations have become increasingly stringent, and the process of inspecting and analyzing individual metal component materials is now a primary concern– particularly to the petroleum and petrochemical refining industry. Today’s best practices include testing 100% of critical materials as part of a QA/QC program.
Elemental analysis of piping and equipment with portable X-ray fluorescence (XRF) technology helps ensure quality assurance of selected weld materials at receiving inspection, in-process welding/fabrication and final inspection check points. A handheld XRF analyzer in the field can help rapidly verify alloys in seconds, recover lost material traceability, and isolate finished welds to validate filler material composition and dilution rates, as well as confirm the integrity of process piping, valves, and reaction vessels.  This analysis could help prevent accidents at the facilities.
In today’s article, we will show an example of PMI testing of a 347 stainless steel FCAW (Flux Core Arc Welding) spool in the as-delivered condition (top photo) vs. the as-welded condition (bottom photo). FCAW is an automatic or semi-automatic welding process using a tubular wire that is filled with a flux throughout its inner core. The flux — which is a chemical cleaning agent, flowing agent, or purifying agent that removes oxidation from the metals to be joined — provides the necessary protection from atmospheric conditions by producing shielding gases during weld metal deposition.
PMI testing was performed for the same heat lot of 347 stainless FCAW directly on the spool before welding followed by the actual weld deposit itself.  Test results yield the absence of Niobium in the first illustration followed by the presence of Niobium in the second illustration and the corresponding alloy grade identification.
In a competitive industrial climate, manufacturers must be vigilant about maintaining the quality of their products.  Even the smallest deviation from specifications of items like raw materials, fasteners, and welds can affect the quality of the product and the reputation of the company. With a handheld XRF analyzer, QA/QC officers can rest assured that no incorrect or out-of-specification metal alloys caused by material mix-ups, lost traceability, incorrect weld chemistry and dilution, or even counterfeit materials enter the manufacturing process.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
15
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Brian Wilson
Business Development Manager, Energy Markets
Field and Safety Instruments
Thermo Fisher Scientific
Comments
  1. Sam Fisher says
    October 12, 2015 at 5:59 pm
    It’s amazing that in today’s world of how much metal we use. Everything has some amount of metal in it. I think that the metals that we use for buildings should be tested more. You never know how many impurities are going to be left over from the melting process.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Weld Metal Analysis: Fine Tune Your PMI Protocol
Vancouver Welcomes ISRI 2015
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook