  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / SEM/EDS Analysis Verifies Difficult Magnet Materials for the Automotive Industry
SEM/EDS Analysis Verifies Difficult Magnet Materials for the Automotive Industry
Written by Carl D. Millholland | Published: 02.16.2016
auto magnetTypical automobiles today use a large number of permanent magnets in motors and sensors throughout a car. For the most part, these magnets are comprised of Neodymium (Nd), Iron (Fe) and Boron (B). However, additions of small amounts of other elements change the magnetic properties of the material as well as its corrosion properties, and in certain cases improve utility at higher temperatures.
As part of incoming inspection, companies that build motors need to confirm that the correct grade of material was supplied or whether the supplied material satisfied the requirements of the grade. In some cases it may be important to investigate failures.
The nature of the material makes this a difficult analysis. NdFeB magnets are formed by sintering a mixture of the powdered constituents at certain temperatures and pressures. The result is a dominant matrix populated by at least two other phases. The exact composition depends on the grade of the material. Heterogeneous structures like this are difficult to analyze.
To obtain the average composition with high accuracy, a typical procedure includes X-Ray Fluorescence (XRF) technology or Inductively Coupled Plasma Mass Spectroscopy (ICP-MS). These techniques produce a very high precision analytical result describing the average composition of the material, but lack any structural information–which is critically important as it is the size, distribution, and composition of the phases that, in part, determine the magnetic properties of the material.
Although heterogeneous materials cannot easily be quantified, they may be characterized. It is straightforward to inspect the material and analyze anomalies. Also, it may be possible to know that two materials are either very similar or different even if the materials cannot be quantitatively described. Finally, it is possible to measure the microstructure in a reproducible manner.
The SEM uses a fine electron beam to probe the sample. The EDS is able to identify the elements present wherever the electron beam impinges on the sample. The electron beam can be focused very tightly such that features much less than one micron across can be inspected. Moreover, little sample preparation is required beyond polishing the sample to a flat surface. For magnets, one additional requirement is that they be demagnetized.  SEM/EDS provides multiple techniques for analysis:
Point Analysis: One of the simplest EDS techniques is to visually inspect a sample and direct the electron beam to conduct analyses at a specific points. The result is one or more elemental analyses as well as an image documenting their locations and the appearance of the sample.
Mapping: A more sophisticated analysis technique is called Spectral Imaging (SI). In this case the field of view is selected by the analyst then the computer rasters the electron beam in a grid pattern across the sample. The result is a two dimensional array of spectra that describe the elemental composition of the area.
Match: Match provides a simple, reliable method for identifying materials based on the similarity of their measured spectra to spectra from known materials.
Feature Sizing and Chemical Typing: FSCT can be used to count the different visible features in a field of view and measure their sizes, shapes and elemental compositions. Most often this is used with Analysis Automation to measure features over a large number of fields. The result is a table of numbers characterizing hundreds, if not thousands of features in a material.
Our next article will outline spectral imaging and mapping of metals in the automotive industry .  If you can’t wait, download SEM/EDS Techniques for Analyzing NdFeB Magnet Material in the Automotive Industry for a detailed explanation of each measurement technique as well as images, spectra, maps showing elemental distributions in a region of the sample, and data tables. 
Share this article
7
SHARES
FacebookLinkedinTwitterMail
gold metallic number 5
5 Precious Metals Analysis Methods
We’ve previously written about the four primary precious m... by Juan Pablo Zuluaga / 05.15.2025
Read More
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Hearts Kept Ticking with Titanium
Spectral Imaging and Mapping Metals in the Automotive Industry
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook