  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / Sheet Metal Thickness Gauge Standards
Sheet Metal Thickness Gauge Standards
Written by Chris Burnett | Published: 10.17.2017
gauging sheet metal thicknessInternational organizations like the ASTM, Japanese Standards Association and others provide guidance on not only alloy chemistry tolerances, but sheet dimensional tolerances and mechanical properties as well. For instrument suppliers, the International Electrotechnical Commission (IEC) has produced standards to provide clear definitions for specific terms and guidance on testing methods used. The IEC standards act as a consistent scale to compare performance from one instrument to another without confusing nomenclature obscuring the true capability of each.
The Nuclear Instrumentation Technical Committee (IEC Technical Committee 45) produced IEC 61336 “Thickness measurement systems utilizing ionizing radiation – Definitions and test methods.” The first committee release was in 1983, and an update was drafted in 1996. The Standard first defines common terms in order to clarify what specific words mean. For example the “mean response time” may be called the “first time constant” by some, and something else by others
The fixed definitions avoid any ambiguity in interpretation. They additionally provide guidance in setting up and carrying out the tests defined in the second part of the standard. The standard dedicates no less than nine terms to clarify time based parameters. Some are related to defining the time associated with the gauge system respond to a change in product thickness, while others are related to the digital processing of signals. This is particularly critical with the advent of high speed data processors. Incoming data can be manipulated and processed by advanced filters to mask or hide true statistical variations.
Radiation measurements are statistical by their very nature. All noise figures should be quoted with a reference to the number of Sigmas, or confidence levels (CL). Most gauge manufacturers present 2 sigma (95% CL) noise figures, but not all. Straightforward data processing provides for predictable results and a better representation of the process dynamics. If a change occurs in the process, advanced filtering may portray a portion of the change, but not the full change. Process engineers and their AGC algorithms may over react, or under correct in response to the manipulated data, thus resulting in poor product quality
In order to ethically improve the speed of the sensor response to change, without increasing the statistical noise on the measurement, the physical characteristics of the sensor need to be optimized for the application. The raw signal; must be shielded to remove as much electrical noise as possible. In an ideal instrument, the radiation detector output is digitized right away. The analog detector signal is converted to a digital number within a few millimeters of its origin. This practically eliminates the possibility of electrical noise impinging on the signal. In comparison tests, following the IEC 61336 guide, the noise on the new designed detector improved by a factor of 30%. Additionally, users of this type of system can benefit further by taking advantage of the detector’s ability to operate at a 1ms mean response time. At this speed, and with the reduced noise, process engineers have the tools to analyze data at high speeds, revealing mill chatter and other higher frequency anomalies.
In a typical rolling mill that produces sheet steel at 250 um, the noise at a 10 ms mean response time might be on the order of +/-0.10% (2 sigma). With the improved signal processing of this system the noise will drop to +/-0.07% (2-sigma). For a mill that produces 2,000,000 tons a year, that translates to a savings of almost $360,000 in raw material alone (using HRC price at $600USD per ton).
An additional benefit to digitizing the signal so early in its journey to the Automatic Gauge Control (AGC) system is speed. Once digitized, the data can be processed without the time consuming ADC / DAC conversions. The reduction of a few milliseconds of process delay time can assure the AGC has time to fully correct any strip thickness deviations. This can result in higher quality product.
Want more details, including graphical representations, charts, and data?  Read the white paper, Optimization of a Cold Rolling Mill with a High Speed X-ray Thickness Gauge.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
196
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane, Editor / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Our Top 5 Gold and Platinum Articles
Portable XRF Analyzers Help Bring New Life to Old Slag
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook