  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Slag: From By-product to Valuable Construction Material
Slag: From By-product to Valuable Construction Material
Written by Stephen Dietz | Published: 07.29.2014
smeltingSlag is a by-product of the metallurgical smelting process. Slag has a double role: it permits removal of impurities, known as gangue, from the melt by forming oxides. It also allows exchange reactions with the liquid metal, permitting control of the process in order for the desirable elements to stay in the melt while the others are removed. (The ore beneficiation process at a mine is designed to remove as much waste and penalty elements as possible prior to transport and smelting.)
Slag is composed primarily of silica and lime, with concentrations of other minor constituents, including alumina, magnesia, and other compounds. During the fluxing process, slag removes the non-ferrous constituents from the molten iron, which purifies the product. Magnesium (Mg) is typically found in concentrations ranging from 1% to 10%, aluminum (Al) from 0.5% to 25%, and silicon (Si) from 5% to 40%. Slag chemistry must be controlled closely to maximize furnace efficiency, reduce impurities in the product, and prolong the life of the refractory lining of the furnace.
Slag is generated in large quantities and has the potential to become a significant source of landfill waste and pollution. But slag itself has value, and technologies have emerged to recycle and reuse reprocessed, granulated slag in different building materials, such as cement, brick, concrete aggregates, wall materials, and glass-ceramic tiles. According to the National Slag Association website, common uses for slag include:
  • Aggregate in granular base, embankments, engineered fill, highway shoulders, and hot mix asphalt pavement
  • Concrete aggregate in bridge construction and in concrete masonry
  • Agricultural applications to increase crop yield
  • Residential and commercial insulation, pipe and process insulation, insulation for ships, mobile homes, and domestic cooking appliances
  • Erosion control, mineral filter, bank reinforcement, and construction of dikes, reefs and sea walls
The goals of slag recycling efforts are both environmental and economic. Reprocessed slag can replace other more costly ingredients used to make building materials. Recycling slag minimizes waste and disposal costs, reduces energy use, extends furnace life, and lowers the amount of additives needed to make slag.
The concerns with slag recycling are that first, because slag can contain environmentally hazardous materials, it must be analyzed for elemental composition before reuse. Second, care must be taken to improve slag potential for other purposes without compromising the quality of the metal is it originally used to produce. Better slag makes for a better finished product, so quick and reproducible analyses of all the oxides in the slag are needed to control slag quality as well as alloy quality during the smelting process.
Compositional analysis of slag by energy dispersive x-ray fluorescence (EDXRF) spectrometry is an ideal way to monitor composition during production to make sure it performs as needed and to evaluate its worth following production. Traditionally, this analysis was done by taking a sample from the furnace (or ladle), then cooling, grinding, and preparing it as a pellet. The pellet was then analyzed using a bench-top EDXRF (or wavelength dispersive – WDXRF) system.
Today, handheld XRF systems are also a good option for slag analysis. These portable analyzers can achieve bench-top grade analytical accuracy and precision closer to the furnace where users can perform testing at lower costs, save time, and improve melt shop productivity.
See study results comparing handheld XRF analysis with study data for slag composition analysis.
Share this article
66
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Microscopy, Microanalysis and Metallographic Society Annual Meeting
VIDEO: XRF Provides Peace of Mind for Vegas Fastener Manufacturing
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook