  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / Titanium: This Lightweight Does Some Heavy Lifting in the Metals Industry
Titanium: This Lightweight Does Some Heavy Lifting in the Metals Industry
Written by Stephen Dietz | Published: 06.11.2014
Titanium racket
Versatile titanium (Ti) can be found everywhere, from tennis rackets to jet engines.
Versatile titanium (Ti) can be found everywhere, from tennis rackets to jet engines. Titanium alloys are used in planes, armor plating, naval ships, spacecraft, and missiles because of their high tensile strength, light weight, extraordinary corrosion resistance, and ability to withstand extreme temperatures. Welded titanium pipe is used in the chemical industry for its corrosion resistance, and its use is growing in petroleum drilling because of its strength, light weight and corrosion resistance. Titanium alloyed with vanadium is used in the outer skin of aircraft, fire walls, landing gear and hydraulic tubing.
Other uses:
  • Due to excellent resistance to sea water, titanium is used to make propeller shafts and rigging and in the heat exchangers of desalination plants and in heater-chillers for salt water aquariums.
  • Because it is considered to be physiologically inert, titanium is used in joint replacement implants such as hip ball and sockets and to make medical equipment and in pipe/tank lining in food processing. Titanium is also used for surgical instruments.
  • Titanium alloys make highly durable and long lasting eye glass frames. Both traditional alloys and shape memory alloys find use in this application.
  • Many backpackers use titanium equipment, including cookware, eating utensils, lanterns and tent stakes because titanium products can be significantly lighter without compromising strength.
  • Titanium is popular in consumer products such as tennis rackets, golf clubs, bicycles, laboratory equipment and laptop computers.
Learn more about the many properties and applications of titanium in the International Titanium Association’s Ti Facts.
The market for titanium is sizeable. According to the U.S. Geological Survey, the value of titanium mineral concentrates consumed in the United States in 2013 was about $1,140 million. About 95% of mined and synthetic titanium mineral concentrates was used to make titanium dioxide (TiO2) a pigment used to impart whiteness to paper and inks. The remaining 5% was used in welding-rod coatings and for manufacturing carbides, chemicals, and metal.
Of the titanium used to make metal, titanium is added in steel alloys to reduce grain size and as a deoxidizer, and in stainless steel to reduce carbon content. Titanium is often alloyed with aluminum (to refine grain size), vanadium, copper (to harden), iron, manganese, molybdenum, and other metals.
The quality and composition of all of the materials that go into making these metal blends must be verified. For example, if the titanium used in the manufacture of propeller shafts or other parts designed for underwater use is not alloyed with the right metal for the application, the parts won’t have the needed corrosion resistance. Similarly, titanium must be alloyed with precise amounts of platinum, palladium, or ruthenium to make pipes that can withstand harsh chemical processing environments.
With both examples, it’s clear that out-of-specification metal alloys can lead to expensive and potentially dangerous part failures. Portable XRF analyzers are indispensible tools for performing material verification for quality control of incoming raw materials, work in progress, and final quality assurance of finished parts, distinguishing alloy grades that are nearly identical in composition to one another in seconds.
Share this article
10
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Priya says
    April 11, 2020 at 11:43 am
    Yes, Titanium Is The Best Metal
    We Deep Steel Alloys Are Stockist Of Titanium Sheet And Plate And Other Raw Materials Like Pipe, Plate, Flange, Fittings, Rod, Coil, Bar, Round Bar, Wire, Electrodes
    Reply
Trackbacks
  1. New Research on 3D Printing with Titanium - Analyzing Metals says:
    May 19, 2016 at 12:33 pm
    […] for its corrosion resistance. For more information about titanium and its many uses, read Titanium: This Lightweight Does Some Heavy Lifting in the Metals Industry or take a look at this infographic to learn 9 Fast Facts About […]
    Reply
  2. Mining Titanium for 3D Printing says:
    May 19, 2016 at 12:46 pm
    […] Discovered by William Gregor in 1791, titanium is a white metal when pure, and is the 9th most abundant element in the Earth’s crust. As a metal, titanium is well known for corrosion resistance and for its light weight but high strength-to-weight ratio. These are probably the reasons it can be found almost anywhere. (See Titanium: This Lightweight Does Some Heavy Lifting in the Metals Industry.) […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
VIDEO: OES Provides Reliable Metal Analysis for Contract Laboratory
Old Ships Find New Life as Scrap Metal
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook