  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / Quality of Galvanized Steel Might Depend on Zinc Coating Weight
Quality of Galvanized Steel Might Depend on Zinc Coating Weight
Written by Chris Burnett | Published: 05.01.2014
Hot Dip Galvanizing (HDG) Line
Hot Dip Galvanizing (HDG) Line with Cold Gauge and Hot Gauge Measurements
Zinc (Zn) is a natural component of the earth and is present in rock, soil, air, and water.  It’s a vital element for the human body, but it’s also one of the most widely-used industrial metals. In the manufacturing world, zinc is used to coat steel, which is used in construction and in numerous consumer products including cars and home appliances. (View this infographic to learn more interesting facts about zinc.)
Zinc coating is the most effective and widely used material to prevent corrosion, maintain the strength, and increase the longevity and performance of the steel. Zinc coating is applied to steel by hot dip galvanizing (HDG), a process by which the steel is passed through a bath of molten liquid zinc or zinc alloy resulting in a protective layer. This galvanized steel is produced at a worldwide rate of nearly 100 million tons each year, and demand for high-quality products is strong. HDG is one of the best ways to produce superior, homogenously coated products in a cost-effective manner.
The final quality of galvanized steel strip depends on various aspects such as material surface, zinc adhesion, and uniform coating weight.  Proper substrate cleaning and strip temperature control govern the first two parameters, but the third, coating weight, is often measured too late in the process to correct. In fact, many producers verify hot dip zinc coating weight after the coil exits the mill.  At this point, if the results are unacceptable the entire coil must be downgraded or scrapped — causing delays, wasted raw materials, and expense.
For HDG lines, automatic coating weight control is essential for uniform production with minimized zinc consumption. The solution to getting the exact weight measurement of zinc coating on steel strip is to measure it during the production process rather than after. X-ray fluorescence (XRF) is an excellent technology to achieve the precision required for such measurements without costly and lengthy delays in production. Online metal coating weight gauges based on XRF technology have proven successful for non-contact coating weight measurement configurable for nearly every type of metal coated product, especially for total zinc coating weight measurements on the difficult-to-measure galvanneal coated sheet.
The measuring units of these gauges use x-ray or gamma ray generators as the source of radiation for coating weight measurement. These sources provide fast response times with all the specific advantages of x-ray generation. The photons from the x-ray source cause XRF radiation to be generated in the coated strip. The XRF radiation is detected by application specific detectors mounted within the measuring unit. With the use of specially designed filters, selective coating weight measurement of various metal layers on the strip is possible.
A critical factor to successful measurement using an XRF-based online coating weight gauge is to create custom calibration standards based on the user’s individual coating processes. Each line should provide all samples required for calibration of their coating weight gauge and for verification of its performance.
Another important factor in obtaining accurate measurement is the placement of the sensors on the coating gauge. A recent white paper, The Benefits of “Hot” Coating Weight Measurements to Coating Control, examines the improvement gained by placing the sensors on the “hot” end of the hot dip galvanizing line, rather than the typical “cold” end. A measuring system placed at the cold end of the hot dip galvanizing line is about 120 meters behind the air knife. In a hot coating weight gauge, the measuring system is only two meters behind the air knife, with a dead time of only about one second. The auto control performance increases significantly in a hot system because the sensors can react to process parameter changes in near-real time, eliminating costly mistakes before they happen and saving on raw materials (zinc). If the measuring equipment consists of both a cold gauge and a hot gauge, the autocontrol can run in cascaded mode. In this case, the cold gauge provides the reference input value for the coating weight set points of the hot gauge. Incorporating coating thickness data from hot and cold sensors, this architecture evaluates long-term modifications in production, and automatically adapts the behavior of the controller to new production conditions.
Read this paper to learn about the ideal architecture for a coating weight measurement and control system which continuously improves its internal control algorithms by comparing the predicted coating weight with measured values.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Share this article
106
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Trackbacks
  1. Our Top 10 Analyzing Metals Articles - Analyzing Metals says:
    June 28, 2016 at 12:45 pm
    […] Quality of Galvanized Steel Might Depend on Zinc Coating Weight Zinc coating is the most effective and widely used material to prevent corrosion, maintain the strength, and increase the longevity and performance of the steel. Zinc coating is applied to steel by hot dip galvanizing. Read the article and access the white paper to learn about the ideal architecture for a coating weight measurement and control system which continuously improves its internal control algorithms by comparing the predicted coating weight with measured values.  […]
    Reply
  2. Online Measurement of Oil Coatings Improves Steel Component Manufacturing - Analyzing Metals says:
    August 4, 2016 at 12:00 pm
    […] can measure the coating without the influence of naturally occurring localized variations in zinc coatings, for example. As the light intensity varies across the spangle, the two-wavelength IR sensor would […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Attendees of Steel – AISTech2014
The High Price of Scrap Metal: A Look at New Efforts to Combat Metal Theft
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook