  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / VIDEO: Can You Dissolve Gold?
VIDEO: Can You Dissolve Gold?
Written by Marlene Gasdia-Cochrane | Published: 05.08.2014
Dissolving gold
Dissolving gold
The University of Nottingham published a video showing gold dissolve into aqua regia (a mixture of nitric acid and hydrochloric acid).  Aqua Regia is a mixture of two acids, hydrochloric acid and nitric acid. Now the exciting thing about Aqua Regia is that neither of the two acids by themselves can dissolve gold. Nitric acid doesn’t really react with gold very much at all, and HCl, hydrogen chloric acid can’t get a hold of the gold to get the reaction going. But if you can put gold into solution that contains both acids, then it can react, and in fact, can indeed dissolve gold – which is why Aqua Regia is also known as ‘royal water.”
So, if you are thinking of getting some gold jewelry for your mom for Mother’s Day this weekend, you can treat her royally, but stay away from the aqua regia.
Watch the video of dissolving gold.
Share this article
375
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Calla Gold says
    May 26, 2014 at 9:54 am
    That is so interesting. I love the story of the gold nobel prizes being dissolved in solution during the war and reconstituted after the war.
    Reply
    • nitonxrf says
      May 27, 2014 at 9:34 am
      Thanks for your comment, Calla.j
      Reply
  2. Ashley DeVille says
    May 29, 2014 at 12:04 am
    Very interesting. Calla, did you discover how they reconstituted the gold? Now that would be interesting.
    Reply
  3. JP BOKOFF says
    January 17, 2020 at 10:34 pm
    I’ve been a pawnbroker for 20 years and didn’t realize adding Nitric and hydrochloric acids together would dissolve gold how would you reclaim it just out of curiosity
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      January 27, 2020 at 2:18 pm
      We’re not in the business of gold extraction, but maybe some of our readers can help you.
      Reply
    • Mattia Siviero says
      July 26, 2020 at 9:18 pm
      you neutralize the acid then you add a metal that’s easier to oxidize zinc is the easiest. Iron is also used. A redox reaction take place where gold oxide is reduced to metallic gold, which precipitates
      Reply
  4. Bill says
    December 3, 2020 at 3:11 pm
    you would add sodium meta bisulfite (Bondie’s Stump out) to the aqua regia mix…To reclaim the gold from solution. One needs to be very careful to only use minimal amounts of acids to dissolve the gold in, and once it is disolved, then you would dilute the acids with water x4..so if you had 100ml acids, then would need minimum of 400-500 ml of water added to acids combined with the stump out…Simply add spoonful of stumpout at a time until no more reaction occurs and begin to see dark colors forming in acid water solution, and allow to settle for hr or few hrs,then filter off the water acid solution and collect powdered gold/brown substance from bottom of container..this is your pure gold of around 995-999 fine gold…rinse well and melt down to gold button,burning filter and all at once.
    Reply
    • Col says
      February 7, 2021 at 4:26 am
      Hi Bill have tried a mix of fine sand with gold particles from creek and added the aqua Regia. Had the reaction as described and then drained the solution out of the mix. Treated the solution with the sub and did not get any gold forming in the bottom of the glass container. is there anything you can suggest I can do. Thanks Col
      Reply
      • Marlene Gasdia-Cochrane, Editor says
        February 8, 2021 at 2:02 pm
        Thanks for your inquiry, Bill. Unfortunately we were just commenting on someone’s video. But maybe our readers can help?
        Reply
  5. Charles Boisseau says
    February 5, 2021 at 2:19 pm
    I’m doing some research. Why would someone dissolve gold? Or gold for that matter. Yes, I heard the story of the scientists who dissolved the Nobel Prizes. But practically, why would someone want to essentially destroy precious metals. I know there are uses and reasons for this that my Google searches are not showing up the mineral experts and rock heads are too smart for me; I need elementary explanation. : -)
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      February 8, 2021 at 2:04 pm
      Thank you Charles for your comment. Let’s hear from our readers!
      Reply
      • Aaron K says
        June 12, 2021 at 6:44 am
        You can dissolve gold to separate and refine it from other metals or impurities. I use aqua regia solution to reclaim gold from scrap. It’s not an easy process, but after a lot of research, one can become a gold refiner with a lot of practice. I run at least one batch of gold chloride every week. Got a nice little collection of gold going.
        Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
The High Price of Scrap Metal: A Look at New Efforts to Combat Metal Theft
New Steel Grades Drive PMI Programs in Automotive Manufacturing
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook