  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / What are Precious Metals and Precious Metals Alloys?
What are Precious Metals and Precious Metals Alloys?
Written by Juan Pablo Zuluaga | Published: 09.29.2020
What are Precious Metals and Precious Metals Alloys?Precious metals are elements that are considered rare and chemically inert. They’re not abundant in nature, and therefore they have high economic value. Precious metals are used commonly in jewelry, industrial processes, or very often as investment vehicles.
The four primary precious metals are gold, silver, platinum, and palladium. Depending on the application, there are common alloying elements that can be mixed with the main precious metals to improve the properties of the final product. There are other elements of interest are that are also used in non-precious metal jewelry or accessories.
When buying, or selling precious metals, one should be familiar with three main terms describing the precious metal:  karat, fineness, and troy ounce.
  • Karat is a grading system to determine the purity of gold. 24 karat is pure gold, so one karat is equal to 1/24th of the pure gold in the alloy. If you do the math, one karat is about 4.16% gold, 14 karats is about 58.3%, and 18 karat is 75% gold, and so on.
  • Fineness is the proportion of the pure precious metal in the alloy. This is often expressed in parts per thousand. If you look closely at a gold bar, sometimes you will see the number 999, which indicates the fineness. It means the bar is 99.9% pure.
  • Troy ounce is the measure to determine the weight of the precious metals. This is a very old term that dates to the middle ages. A troy ounce is about 31.1 grams, as opposed to a regular ounce, which is about 28.35.
It is common to find five different types of gold on the market:
  1. Fine gold, which is basically pure gold with fineness of 999 or above.
  2. Gold alloy is simply gold that has been mixed with other elements.
  3. Yellow gold contains silver as the main alloying element.
  4. Rose gold contains copper as the main alloying element.
  5. White gold contains either palladium or nickel as the main alloying elements.
Just like in any other businesses such as pawn brokers, cash-for-gold operations, jewelry stores and gold traders, we encounter some risks and challenges. One risk involves the material. There is a possibility of overpaying the seller because you cannot determine the correct karats or because you think it’s really solid gold when, in reality, it’s a plated material. Another challenge is determining what other precious metals besides gold are present in your sample. Sometimes you have other precious metals that are valuable, like palladium or platinum, and you can’t really see them just by looking at the piece. Sometimes the piece can contain non-precious metals such as nickel, or even some toxic metals like lead.
Verification tests that business use to combat these risks include the scratch and acid test, which is widely used but not very accurate and potentially dangerous. The most precise method is fire assay. However, this method destroys the sample. Laboratory methods with expensive machines require extensive sample preparation. Finally, there is portable X-ray florescence (XRF), a non-destructive method that measures up to 22 elements in one reading.  Therefore, if you are a businesses that needs an accurate, fast, easy, non-destructive analysis method for a reasonable price, portable XRF analyzers are one of the best possible solutions among all the techniques.
We’ve gone into more details about analyzing and verifying precious metals and precious metal alloys in our webinar Not All that Glitters is Gold. You can download the recording anytime and see a demo of the analysis in action.
  • Watch the recording: Not All that Glitters is Gold.
Share this article
248
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. eBullion Pvt Ltd. says
    November 21, 2023 at 9:02 am
    Exploring the world of precious metals and alloys opens up a treasure trove of insights. The blog delves deep into the significance and applications, making it a must-read for anyone curious about these valuable elements.
    Reply
  2. John says
    December 25, 2024 at 9:53 am
    Great breakdown of precious metals and their alloys! The article provides valuable insights into their properties, uses, and the importance of accurate testing methods. It’s a must-read for anyone looking to deepen their understanding of these valuable materials.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
ebook: A Practical Guide to Metals Gauging Systems
5 Precious Metals Analysis Methods
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook