  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / White Gold vs. Platinum: Can You See the Difference?
White Gold vs. Platinum: Can You See the Difference?
Written by Jonathan Margalit, PhD | Published: 06.03.2014
Wedding RingsAccording to “The Knot” — a popular wedding planning website — the most popular month to marry in the US is June. And what’s the most popular metal used for wedding rings?  Well, although there are several kinds of precious metals, most couples choose gold or platinum.  What’s the difference between the two metals?
Gold and platinum are both dense, malleable, ductile precious metals used in jewelry and industrial applications. Throughout history, gold has always been prized as a symbol of wealth, but platinum is, currently, more valuable than gold. There are many other important differences between these two elements.
Let’s start with the better-known gold. Gold is represented by the symbol Au on the periodic table of elements. Gold is appropriate for some industrial applications because it doesn’t rust or corrode and it conducts both heat and electricity, but gold is valued mostly as jewelry or coins.
As jewelry, gold is typically yellow, white, or red, depending on the alloying elements. White gold doesn’t occur naturally; it gets its silvery hue by being alloyed with white metals such as silver, nickel, manganese, platinum, or palladium. For a shiny finish, white gold is usually coated in rhodium plating that must be replaced periodically to prevent the white gold from reverting back to yellow.
In the industrial world, gold is used primarily as connectors in electronic devices, such as cell phones and computers, because it’s corrosion and tarnish resistant, conducts electricity well, and disperses heat quickly. Various amounts of gold can be found in some large appliances such as microwaves, washing machines, and TVs.
Now let’s look at platinum, represented by the symbol Pt on the periodic table. Platinum is also used to make jewelry and looks similar to white gold, but when compared side by side with white gold you can see that platinum has a more grayish color.  Platinum doesn’t fade to yellow like white gold but the finish will dull, so like white gold, platinum jewelry may require some maintenance to keep it shiny.
Platinum is more dense than gold, and by law jewelry that is labeled “platinum” must be at least 95% pure platinum, so a platinum ring will weigh, and therefore cost, more that a gold ring of the same size. (14k gold is only 58.5% gold and 18k gold is 75% gold). Platinum is stronger and more durable than gold. However, a distinct advantage of platinum is that it’s hypoallergenic, whereas some people will have an allergic reaction to white gold due to the nickel alloy it contains. Palladium, one of the Platinum Group Metals, may be an even better choice than platinum because it has similar characteristics as platinum at a lesser price.
Platinum and palladium may be good choices for jewelry, but most of these metals are produced for industrial purposes such as catalytic converters and numerous electronic components. Platinum can also be used in diesel engines, and petroleum and chemical refining catalysts. Palladium is used to make metallic glass, polymer membranes, coatings, plastics, textiles, and fuel cells. Palladium nanoparticles are finding applications as potential drug delivery mechanisms.
Market analysts project continued volatility in the price of gold while the price of platinum is expected to rise due to dwindling supply and increasing demand. For businesses that buy and recycle precious metals such as jewelry stores, refineries,  and pawn shops, failing to distinguish between white gold, platinum, and palladium or to detect even a small variation in the composition of one of these precious metals can be an expensive mistake.
Many businesses are now turning to X-ray fluorescence (XRF) technology for fast, accurate, and most importantly, nondestructive analysis of the precious metals that come into their shops. XRF quickly provides the exact karat weight and percentages of all elements within an item – easily identifying non-standard, under-karated, and even advanced counterfeit material.
Share this article
264
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. kanishk says
    October 30, 2020 at 8:08 am
    There are very minor differences one can notice. some of them are the lustre in some case as well as the dents. platinum are metals tat cant endure dents much and is very difficult to bring them in their original form once dented. however that’s not the case with white gold. I must say you guys have curated some good content. Keep up the good work!
    Reply
Trackbacks
  1. Our Top 10 Analyzing Metals Articles - Analyzing Metals says:
    June 28, 2016 at 12:01 pm
    […] White Gold vs. Platinum: Can You See the Difference?Gold and platinum are both dense, malleable, ductile precious metals used in jewelry and industrial applications. Many businesses are now turning to X-ray fluorescence (XRF) technology for fast, accurate, and most importantly, nondestructive analysis of the precious metals that come into their shops. XRF quickly provides the exact karat weight and percentages of all elements within an item – easily identifying non-standard, under-karated, and even advanced counterfeit material. Read more. […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Robotics-based Automation Advances Steel Production Quality Control
VIDEO: OES Provides Reliable Metal Analysis for Contract Laboratory
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook