  • Contact
Accelerating ScienceAnalyzing Metals / General/Industry / Wood at One-Fifth its Original Thickness Outperforms Steel and Titanium
Wood at One-Fifth its Original Thickness Outperforms Steel and Titanium
Written by Chris Burnett | Published: 04.17.2018
Wood at One-Fifth its Original Thickness Outperforms Steel and TitaniumCould treated wood replace steel and titanium? Engineers at the University of Maryland in College Park say they have developed a way to make wood strong enough to give titanium alloy a run for its money.
“This new way to treat wood makes it twelve times stronger than natural wood and ten times tougher,” said Liangbing Hu, research team leader. “This could be a competitor to steel or even titanium alloys, it is so strong and durable. It’s also comparable to carbon fiber, but much less expensive.”
“It is both strong and tough, which is a combination not usually found in nature,” said Teng Li, the co-leader of the team. “It is as strong as steel, but six times lighter. It takes 10 times more energy to fracture than natural wood. It can even be bent and molded at the beginning of the process.” 
The process involves removing the wood’s lignin, then compressing the wood under mild heat, causing the cellulose fibers to become so tightly packed that they form strong hydrogen bonds. The compression makes the wood five times thinner than its original size.
“This kind of wood could be used in cars, airplanes, buildings – any application where steel is used,” Hu said.
The study was published in the journal Nature.
Titanium is highly valued in the metals industry for its high tensile strength, as well as its light weight, corrosion resistance, and ability to withstand extreme temperatures. It’s as strong as steel but 45% lighter, and twice as strong as aluminum but only 60% heavier. Its ability to withstand extreme temperatures is a real technical advantage for many applications, but it presents serious challenges for titanium sheet producers. Thickness reductions require very high forces; sheet metal thickness measurement tools and on-line thickness gauges are essential for process optimization. As much of a challenge as this material is to roll, you don’t want to do it twice.
Steel is also widely-used for its strength as well as flexibility. It is flexible in the mechanical sense, but also in terms of the wide varieties of applications where it can be used.  There are hundreds of different grades, thicknesses, and strengths of stainless steel, which are created by alloying steel with different elements. Martensitic or semi-austenitic steels are the strongest due to the addition of elements such as aluminum, copper and niobium.
Steel starts out as flat sheet metal or plates and must be manufactured to precise thickness specifications depending on the application for which it is used. It must also be easily machinable so that it can be formed into its permanent shape without cracking. While strength is an advantage in many applications, adding strengthening alloys may contribute poor machinability. Accurate thickness measurement of process-line steel ensures the finished products have specific mechanical properties, including the appropriate strength and stiffness for their application. An excellent way to accomplish this is by processing the material through a cold rolling mill. Cold rolling is a metal forming process in which a sheet of metal is pressed through a pair of rolls to reduce thickness, increase strength, and improve surface finish. Modern cold rolling mills are able to achieve high speed production of sheet steel with the help of an x-ray thickness gauge. These metal x-ray thickness gauges can detect and correct deviations in thickness in real-time to achieve high quality steel strip.
Read Optimization of a Cold Rolling Mill with a High Speed X-ray Thickness Gauge.
Share this article
325
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Rich Kovaly says
    December 10, 2020 at 4:35 am
    this is all fine and dandy, but the whole reason of using steel in the first place is because for one it’s stronger than wood but for two because of deforestation issues, and the simple fact that we need our trees I to survive. You know the whole carbon dioxide oxygen thing?
    If you’re going to go back and start using wood to replace steel again then you’re going back to the old practices of using too much wood, which leads to supply and demand and lear cutting and deforestation again. Which I thought we had been trying to eliminate for the last 20 years
    Reply
  2. Angelo Lubag says
    February 5, 2021 at 11:31 pm
    I see Rich Kovaly’s point clearly. However, if industry and government will see this clearly, mining ores (which destroys the environment) could be minimized. Also, it should trigger the business of reforestation since it could make a money making sense to grow much more wood. Governments should encourage it in that sense. We have so much steel to recycle to reduce mining and so much land that we deforested to plant again for future structures. If businesses will just cut old growth trees, then it wont be sustainable…
    Reply
  3. Angelo Lubag says
    February 5, 2021 at 11:33 pm
    I see Rich Kovaly’s point clearly. Steel cannot be supplanted by this technology for the most part. However, if industry and government will see the advantages of this tech clearly, mining ores (which destroys the environment) could be minimized. Also, it should be used to trigger the business of reforestation since it could make a money making sense to grow much more wood. Governments should encourage it in that sense. We have so much steel to recycle to reduce mining and so much land that we deforested to plant again for future structures. If businesses will just cut old growth trees, then it wont be sustainable…
    Reply
  4. Theocrates Eustacius says
    February 18, 2021 at 6:44 am
    The point is to use fast growing soft species and leave old growth hard woods alone. Although not mentioned, this process can probably be used to transform bamboo, which grows like a weed, so to speak. Because these materials are stronger and lighter than steel, they may be substituted in many uses, such as biodegradable vehicles. It does not require much imagination to realize complex shapes can be molded using progressive dies or complex molds pressed using steam. Next step is to apply 3-D printing technology to these “plasticized” soft wood feedstocks.
    Reply
    • Chris Hale says
      December 2, 2022 at 11:06 pm
      Most wood that is used commercially in the US comes from specially bred trees that grow in a couple years, and they harvest the areas so as not to cut the whole thing down. I’m sure other countries don’t all do that, but the US has become pretty sustainable in the way we do that. If you compare a modern 4×4 to one from 40-50 years ago, you can immediately tell the difference. The modern ones are much lighter and more flimsy, almost like balsa wood in comparison
      Reply
  5. bill says
    November 18, 2021 at 6:01 pm
    If the wood ends up compressed and used in buildings for decades (or more), it is also a good carbon sink. I wonder how much energy this process requires compared to the energy needs for steel?
    Reply
  6. Jamie Thompson says
    December 6, 2022 at 7:34 am
    Very interesting, but is it fire and mold resistant?
    Reply
  7. RCG says
    January 2, 2023 at 9:25 pm
    Wait until major league baseball finds out about this. Routine warning track fly balls will become home runs.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
NACE Corrosion 2018 – World’s Largest Corrosion Conference and Expo
Scrap Aluminum’s Green and Automotive Applications
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook