  • Contact
Accelerating ScienceAnalyzing Metals / Metals & Alloys / Zero Waste Initiatives a Boon for the Scrap Metal Industry
Zero Waste Initiatives a Boon for the Scrap Metal Industry
Written by Stephen Dietz | Published: 07.22.2014
metal recycling zero waste“Zero waste” is becoming a popular term in the recycling business. Many industries have implemented programs to reduce most, if not all, materials normally destined for landfill. So what exactly is zero waste?
The goal of zero waste is to completely eliminate waste rather just reduce it, or burn it, or dump it in a landfill. The Zero Waste International Alliance defines the zero waste philosophy as “…designing and managing products and processes to systematically avoid and eliminate the volume and toxicity of waste and materials, conserve and recover all resources, and not burn or bury them. Implementing Zero Waste will eliminate all discharges to land, water or air that are a threat to planetary, human, animal or plant health.”
On an industrial level, zero waste means repurposing everything that goes into a manufacturing process.  The benefits are both environmental and financial. A fact sheet published by the World Steel Organization describes the organization’s goal of achieving zero waste in the steel industry through the recovery and reuse of steel manufacturing by-products such as slag, sludge, and dust. Recycling by-products reduces landfill waste and CO2 emissions, helps preserve natural resources, and reduces raw material use. Recovered by-products also can be sold to other industries to create an additional revenue stream.
For example, iron ore-based steelmaking generates slag as a by-product which is a potentially significant and possibly toxic source of landfill waste and pollution. But technologies have emerged to recycle and reuse reprocessed, granulated slag in different building materials, such as cement, brick, concrete aggregates, wall materials, and glass-ceramic tiles. Other technologies include pelletizing and briquetting, which involve mixing a variety of steel-making by-products including dusts, sludge, and sinter fines, for reuse in steel production.
The automotive industry is currently the largest producer of scrap metal, so it makes sense that several automotive companies have implemented successful zero waste or “nil to landfill” initiatives to recycle manufacturing materials, including scrap metals. Efforts include reductions in the size of steel sheets used to make car bodies, and recycling scrap from unused parts and machining and trimming processes. One automotive company claims to have recycled more than half a million tons of scrap in the U.S. and Canada alone in one year, worth hundreds of millions in revenue.
According to the U.S. Geological Survey 2014 Iron and Steel Scrap Mineral Commodity Summary, in 2012, the automotive recycling industry recycled more than 18 million tons of steel from end-of-life vehicles through more than 300 car shredders, the equivalent of nearly 18 million automobiles. This scrap continues on through the industrial supply chain, purchased mostly by iron and steel manufacturers to reduce their consumption of raw materials used to produce steel products for the appliance, construction, container, machinery, oil and gas, transportation, and other industries.
With so much scrap metal coming into scrap yards, scrap metal recycling has become big business. Scrap yard operators rely on a variety of tools to sort and prepare the scrap for its various applications. One of these tools is handheld x-ray fluorescence (XRF) analyzers, which are used to determine the composition of the metal so that it can be sorted correctly. Scrap metal recyclers also use handheld XRF analyzers to positively identify numerous alloys at material transfer points and to guarantee the quality of their product to their customers. Elemental analysis of scrap, including the existence of contaminants or hazardous elements, is also needed for quality, safety and regulatory compliance.
Read about one scrap metal company’s experience using an XRF handheld analyzer.
Share this article
5
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Nirmal Group says
    April 9, 2022 at 5:48 am
    Hello there!!! It was wonderful to go through your blog. It is full of information and adds great values to the readers. This kind of blog helps readers like us in many ways and increases our interests. Thank you for sharing and looking forward to reading more of your blogs.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Does Platinum Migrate During Electrode Assembly?
Next-Generation Personal Radiation Detectors Provide a New Level of Protection for Metal Recyclers
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook