  • Contact
Accelerating ScienceAdvancing Mining / General/Industry / 9 Fast Facts About Lutetium: An Infographic
9 Fast Facts About Lutetium: An Infographic
Written by Chris Calam | Published: 07.23.2019
9 Fast Facts About Lutetium: An Infographic  According to the USGS, “The REE (Rare Earth Element) group is composed of 15 elements that range in atomic number from 57 (lanthanum) to 71 (lutetium) on the periodic table of elements, and are officially referred to as the ‘lanthanoids,’ although they are commonly referred to as the ‘lanthanides.’”
This silvery white metal, number of 71 on the periodic table, occurs in very small amounts in nearly all minerals containing yttrium, and is considered a heavy rare earth element.
The U.S. Los Alamos National Laboratory reports that “The pure metal has been isolated only in recent years and is one of the most difficult to prepare. It can be prepared by the reduction of anhydrous LuCl3 or LuF3 by an alkali or alkaline earth metal. The metal is silvery white and relatively stable in air. 176Lu occurs naturally (2.6%) with 175Lu (97.4%). It is radioactive with a half-life of about 3 x 1010 years.”
According to the Laboratory, stable lutetium nuclides, which emit pure beta radiation after thermal neutron activation, can be used as catalysts in cracking, alkylation, hydrogenation, and polymerization. Virtually no other commercial uses have been found yet for lutetium.  It cautions, however, that while lutetium, like other rare-earth metals, is thought to have a low toxicity rating, it should be handled with care until more information is available.
As mentioned, REEs rarely exist in pure form; they are usually concentrated in more than one mineral, and each mineral requires a different costly extraction technology and mineral processing. Geochemical exploration is the main method of REE exploration.
Depending on the REE project type, handheld XRF analyzers are useful instruments that can provide real-time, on-site assays of REEs and other elements in any type of geological samples. They are indispensable tools for the analysis of the light series of REEs (LREEs), including lanthanum (La), cerium (Ce), praseodymium (Pr), and neodymium (Nd). Other elements associated with REE-bearing minerals such as thorium (Th) and Yttriym (Y) can also be analyzed. By using the concentrations from these elements, especially Y, it is possible to infer concentrations of heavy REEs (HREEs), like Lutetium, that are commonly associated with Y-containing host minerals.
Download this infographic and discover 9 Fast Facts About Lutetium
infographic: lutetium
###
Share this article
205
SHARES
FacebookLinkedinTwitterMail
Petrochemical plant illuminated at twilight
Where Do Flowmeter Measurements Factor Into Downstream Oil and Gas?
In the downstream sector of the oil and gas industry—where... by Ethan Borchelt / 05.06.2025
Read More
Large industrial water treatment and boiler room. Shiny steel metal pipes and blue pumps and valves.
How to Better Detect Methane, CO, CO₂, and Volatile Organic Compounds (VOCs) Leaks
The oil and gas industry faces stringent regulatory complian... by Bob Gallagher / 02.18.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
How is the Demand for Iron Ore as it Relates to the Steel Industry?
6 Points in the Production Process to Optimize Cement Making
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook