  • Contact
Accelerating ScienceAdvancing Mining / Laboratory Analysis / Analysis of Ilmenite Ore Samples from Australia and China with XRF and XRD
Analysis of Ilmenite Ore Samples from Australia and China with XRF and XRD
Written by Christopher Shaffer | Published: 03.12.2019
Analysis of Ilmenite Ore Samples from Australia and China with XRF and XRDX-ray diffraction (XRD) and X-ray fluorescence (XRF) are most often used for complimentary qualitative and quantitative analysis. XRD is commonly used in geology and mining (earth science) laboratories to characterize the mineralogical composition, while XRF is routinely used to determine chemical composition. Here we present a case study in which XRD and XRF were used to analyze the mineralogy of ilmenite ore samples from China and Australia and demonstrate clear differences between the deposit locations.
Ilmenite (FeTiO3) is the most important titanium ore, responsible for up to 47% of the total worldwide production of TiO2, a whitening pigment. The world’s largest producer of ilmenite ore is Australia, together with South Africa, China and Canada. The ore occurs as two different morphologies, hard rock and heavy sand ilmenite, whereof the latter is more utilized. Ilmenite forms solid solution series with geikielite (MgTiO3) and pyrophanite (MnTiO3) and can more generally be written as (Fe,Mg,Mn)TiO3.
For evaluation of the grade of the ore, it is crucial to know both the chemical and mineralogical composition of the sample, which are usually analyzed by XRF and XRD respectively.
The ore from Australia only contains expected minerals (valuable raw materials) whereas are samples from China show a wider variety of mineral phases, which are all related to ilmenite deposits.
Read the complete study, including tables and charts illustrating analytical results:
Analysis of ilmenite ore samples from Australia and China with ARL EQUINOX 100 XRD & ARL QUANT’X XRF instruments
Elemental and Mineralogical characterization of geological materials is of primary importance both in terms of assessing their natural resources and in terms of their exploration, beneficiation and extraction processes for industrial needs.
 Additional recommended reading:
Analysis of geological minerals using the ARL EQUINOX series X-ray diffractometer.
Watch this video to see a demonstration of a desktop XRD instrument.
Share this article
FacebookLinkedinTwitterMail
Petrochemical plant illuminated at twilight
Where Do Flowmeter Measurements Factor Into Downstream Oil and Gas?
In the downstream sector of the oil and gas industry—where... by Ethan Borchelt / 05.06.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Oil Or Gas Transportation With Gas Or Pipe Line Valves On Water
Balancing Midstream Hydrocarbon Gathering Systems via Flow Measurement and Computation
In the midstream hydrocarbon industry, maintaining balance w... by Ethan Borchelt / 01.07.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Improving Oil and Gas Exploration with Drones
New Minerals Capture CO2 in Mine Waste
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook