  • Contact
Accelerating ScienceAdvancing Mining / General / Do Mining Companies Need to Annually Re-Calibrate their XRF Analyzers?
Do Mining Companies Need to Annually Re-Calibrate their XRF Analyzers?
Written by Paul Pazareskis | Published: 02.09.2016
niton xrf analyzerOur sister blog, Analyzing Metals, published a useful article last week regarding the importance of maintaining your XRF instrument, including checking calibrations every year, to ensure they will correctly determine the elemental composition of materials. The accuracy of this technology is especially important in mining where you need to maximize your exploration budgets, pre-screen laboratory samples, identify drill targets quickly, reduce re-mobilization costs, and get an accurate report to the capital markets as fast as possible.
XRF analyzers determine the chemistry of a sample by measuring the fluorescent (or secondary) x-ray emitted from a sample when it is excited by a primary x-ray source. Each of the elements present in a sample produces a set of characteristic fluorescent x-rays (“a fingerprint”) that is unique for that specific element, which is why XRF spectroscopy is an excellent technology for qualitative and quantitative analysis of material composition. 
Portable analyzers allow you to take assay analyses in real time, and can be used in mine mapping, ore grade control, and environmental compliance. and the exploration and mining of a wide variety of base metals, precious metals, rare earth elements, mineral fuels, industrial minerals, and gas-bearing strata samples.
But the instrument must be precisely calibrated in order to ensure the results are accurate. Read the article to see why re-calibration is needed on an annual basis. You need your results immediately, so make sure they are accurate as well.
Editor’s Note: If you own a Thermo Scientific Niton XRF portable analyzer, contact 1-800-875-1578 or 1-978-670-7460 to discuss the Annual Calibration & Certification program with the Technical Support team to get your analyzer calibrated and certified.”
Share this article
FacebookLinkedinTwitterMail
Large industrial water treatment and boiler room. Shiny steel metal pipes and blue pumps and valves.
How to Better Detect Methane, CO, CO₂, and Volatile Organic Compounds (VOCs) Leaks
The oil and gas industry faces stringent regulatory complian... by Bob Gallagher / 02.18.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
They Won’t Be Throwing Louisiana Opal Beads at Mardi Gras
Diamond Roses Mined for Valentine’s Day
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook