  • Contact
Accelerating ScienceAdvancing Mining / Elemental/Geochemical Analysis / Fascinating Facts About Manganese
Fascinating Facts About Manganese
Written by Ali Somarin | Published: 05.29.2014
Manganese InfographicIt’s harder and more brittle than iron.  Your body needs it to function properly, but too much can be unsafe.  It’s found in several foods – including nuts, tea, whole grains, and leafy vegetables – as well as in construction equipment, dry cell batteries and plant fertilizers.  It’s Manganese, a gray-white metal, with atomic number 25 on the periodic table. Manganese (Mn) is not a rare metal as it makes up about 0.1% of the earth’s crust. Manganese ores usually consist of dark brown to black oxides particularly pyrolusite (MnO2) and psilomelane [(Ba,H2O)2Mn5O10]. Manganese carbonate (rhodochrosite, MnCO3) and silicate (braunite, MnSiO3) may occur locally. All manganese minerals usually show a close spatial relation to the iron ores. Manganese is mined in countries such as South Africa, Australia, China, Brazil, Gabon, Ukraine, India, Ghana and Kazakhstan.  According to the United States Geological Society, “Land-based manganese resources are large but irregularly distributed; those in the United States are very low grade and have potentially high extraction costs. South Africa accounts for about 75% of the world’s identified manganese resources, and Ukraine accounts for 10%.” The USGS reports that the value of domestic consumption, estimated from foreign trade data, was about $950 million.  Wants some quick facts about Magnesium?  Take a look at this infographic that lists 9 Fast Facts about Manganese. 
Share this article
17
SHARES
FacebookLinkedinTwitterMail
Petrochemical plant illuminated at twilight
Where Do Flowmeter Measurements Factor Into Downstream Oil and Gas?
In the downstream sector of the oil and gas industry—where... by Ethan Borchelt / 05.06.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
A Snapshot of Strategic Metals
Ubiquitous Industrial Minerals: Nature’s Most Popular Raw Materials
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook