  • Contact
Accelerating ScienceAdvancing Mining / General / Forecasted Demand and Supply Gap for Critical Battery Minerals
Forecasted Demand and Supply Gap for Critical Battery Minerals
Written by Ankush Koul | Published: 06.13.2023
Pile of lithium-rich salt material from deposits for Li-Ion battery manufacturing in EV industry, Lithium hexafluorophosphate extract from rechargeable energy cell in recycling process 3D illustrationBattery technology has existed for over two centuries and has evolved significantly and become a crucial modern life component. Batteries have a wide range of uses, from powering small devices like cell phones and laptops to large-scale applications like electric vehicles and renewable energy storage systems.
The growing demand for batteries is driving the need for critical battery minerals like lithium, cobalt, and nickel. According to a report by the International Energy Agency, the demand for these minerals could increase by more than 40 times over the next 20 years. However, the supply of these minerals is limited, and there is a growing concern about the environmental and social impacts of mining.
The significance of battery technology lies in its potential to transform the energy landscape, reducing our reliance on fossil fuels and enabling a more sustainable future. However, as the demand for batteries continues to grow, so does the need for efficient mining of the resources.
According to the USGS Mineral Commodity Summaries 2023 report, here are some facts about three of the critical battery minerals we mentioned above:
Cobalt
Global cobalt mine and refinery production were forecast to increase to record-high levels in 2022. Most cobalt is mined as a byproduct of copper or nickel. China was the world’s leading producer of refined cobalt, most of which was produced from partially refined cobalt imported from Congo (Kinshasa). China was the world’s leading consumer of cobalt, with about 80% of its consumption used by the rechargeable battery industry.
Lithium
Worldwide lithium production (excluding U.S. production) in 2022 increased by 21% to approximately 130,000 tons from 107,000 tons in 2021 in response to strong demand from the lithium-ion battery market and increased prices of lithium. Global consumption of lithium in 2022 was estimated to be 134,000 tons, a 41% increase from 95,000 tons in 2021…. Owing to continuing exploration, identified lithium resources have increased substantially worldwide and total about 98 million tons.
Nickel
Estimated global nickel mine production increased by about 20%, with almost all of the increased production attributed to Indonesia. The largest share of the increase was facilitated by the ongoing commissioning of integrated nickel pig iron and stainless-steel projects. In addition, several companies continued to develop projects to produce intermediate matte or mixed nickel-cobalt hydroxide that were intended to be used as feedstock to produce battery-grade nickel sulfate.
XRF Technology in Mineral Exploration
Analytical technologies can help bridge the gap between the forecasted demand and supply of critical battery minerals. For example, handheld instruments utilizing X-ray fluorescence (XRF) spectroscopy can be used to analyze the composition of mineral deposits, helping to identify the most promising sources of raw materials.
XRF is a non-destructive analytical technique used to determine the elemental composition of materials. Handheld XRF analyzers work by measuring the fluorescent (or secondary) X-rays emitted from a sample when excited by a primary X-ray source. Each of the elements present in a sample produces a set of characteristic fluorescent X-rays, or “unique fingerprints”. These “fingerprints” are distinct for each element, making handheld XRF analysis an excellent tool for quantitative and qualitative measurements.
At the exploration stage, a quick, non-destructive analysis is often required for outcrop and soil analysis, advanced exploration and drilling, core sample analysis, mine mapping, grade control, and cuttings analysis for mud logging and reservoir characterization. This data can enable geologists and miners to make data driven decisions on locations and sites.
Portable XRF analyzers enable miners and geologists to perform fast, accurate geochemical analysis – virtually anywhere on site – and quickly identify and recover the most economically viable resources.
Additionally, advanced data analytics can be used to optimize the extraction process, reducing waste and improving the efficiency of the mining operation.
Society’s plans for a decarbonized world will rely on renewable energy and batteries, which rely on mining the right metal elements. Meeting the demand for cobalt, lithium, nickel, and other clean energy minerals calls for innovations and the adoption of technologies that will help provide fast and accurate geochemical analysis. Portable XRF analyzers help identify drill targets, increase discovery rates, enable on-site decisions, and pinpoint where to focus on the grid – ultimately guiding geochemical decisions and optimizing mining operations.
Share this article
44
SHARES
FacebookLinkedinTwitterMail
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Technology Considerations for Bulk Ore Sorting
Indonesia Miner Conference Focused on Metals
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook