  • Contact
Accelerating ScienceAdvancing Mining / Data Management/Lab Automation / New Software Offers Advantage in Underground Mining
New Software Offers Advantage in Underground Mining
Written by Chris Calam | Published: 12.05.2017
Underground mineFinding productive mine sites begins with surveying potential locations to ensure the area has enough resources to make further exploration worthwhile. Once a site has been identified, mining geologists may use portable X-ray fluorescence (XRF) analyzers to perform ore grade control to figure out where the most profitable ore bodies are in terms of location and mineral concentration variability.
Ore deposits are inconsistent in nature, having high concentrations of metals in one area but much lower concentrations in others. Portable XRF analyzers provide fast acquisition of geochemical data for ore deposit modeling, determining ore boundaries, and quickly identifying increasingly low-grade deposits. The data collected during the grade control processes helps determine metal concentrations and identify areas for drilling or more detailed investigation. This data can be used to help create geochemical maps to see an anomaly or trend, make the most efficient and economical drilling and excavation decisions, and ensure that a mining plan will yield a sufficient return on investment.
An underground mine plan includes mapping out a network of transportation tunnels. Mining surveys can also be used to provide information about mine and tunnel dimensions, which helps determine the feasibility of using the large trucks and equipment needed throughout the mine. Now a unique approach to efficient planning of underground mine tunnels has been developed by the software company MineOptima, founded by researchers at the University of Melbourne. The research team, headed by Professor Doreen Thomas and Professor Hyam Rubinstein,  applied the principle of network optimization to the design and planning of the complicated, underground tunnel networks used by large haulage trucks that bring ore to the surface. An article on the University of Melbourne web site explains their process:
Professor Thomas says that there are complex rules that underground tunnels need to satisfy, such as precise constraints on the gradients of the incline and the decline.
“The gradient constraint is significant because the huge haulage trucks can only drive up a very shallow gradient. Networks need to slope very gently from the ore bodies to the surface. The other constraint is that the large trucks can only turn along wide corners in a tunnel.”
The software takes these restrictions into consideration – plotting the shortest network of paths between the mine’s entrance and the various points around the underground orebody.
The MineOptima software uses detailed geological information obtained from exploratory drilling that maps out the locations of ore bodies and other underground features such as faults and weak ground. The software then uses algorithms developed by the University of Melbourne Network Research Group to explore a range of possibilities for safe and optimal access tunnels, plotting their possible location and the shortest network of paths between the mine’s entrance and the various points around the underground orebody.
Recently, international mining software and consulting firm RPM Global, acquired MineOptima.
Read the full article, Using maths to map mines deep underground.
Share this article
FacebookLinkedinTwitterMail
Engineers and staff communicate on site in chemical plant
Why Flow Computing is Key to Minimizing Disruptions in Station Control
Today’s station control systems—whether in oil and gas, ... by Ethan Borchelt / 04.08.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Li-ion Battery Supply: Can Lithium and Cobalt Meet the Challenge?
Complementary XRF and XRD for More Complete Analysis of Geological Minerals
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook