  • Contact
Accelerating ScienceAdvancing Mining / Oil and Gas / Portable XRF Calibrations in Well Site Geochemistry
Portable XRF Calibrations in Well Site Geochemistry
Written by Esa Nummi | Published: 04.30.2015
Well Site Portable XRF CalibrationsExploration and production (E&P) of unconventional oil and gas requires advanced and innovative technologies. Horizontal drilling and fracking are examples of innovations that opened the door to unconventional hydrocarbon resources. Well site geochemistry is another new technique that can provide invaluable and near real-time data, with practical applications from precise rock identification to mineralogy and mineral modeling, chemostratigraphy, geo-steering, reservoir characterization (permeability, porosity, fracture population, brittleness) and even productivity (TOC inference). This technique is not unknown in E&P and is currently used in chemostratigraphy and mud logging.
The data for well site geochemistry is commonly provided by using laboratory instruments, an often expensive and time-consuming process. A supplemental technique is portable X-ray fluorescence (PXRF), which uses small, portable and rugged handheld or benchtop analyzers. These analyzers can provide timely data on a variety of samples, from drill cuttings to cores and outcrops, without the need for a highly skilled workforce.
3 Calibration Methods
When using PXRF, a common question is, “Do I need to calibrate the analyzer?” There are three calibration methods that are used in PXRF: Empirical, Fundamental Parameter (FP), and Compton Normalization.
Both FP and Compton Normalization calibrations are performed at the factory.
  • Empirical calibration needs known samples (e.g.. standards) and uses the correlation between the readings obtained from these samples and their known values in order to quantify elements in unknown samples. This calibration can provide precise and accurate assay data for a narrow range of sample types and matrices. Unreliable data can be created, however, if the sample type or matrix differs from the ones that were used for calibration.
  • Compton Normalization only works with a sufficient scatter (Compton Peak) in a spectrum related to the sample matrix. As the average atomic mass of the sample increases, the Compton peak decreases. Therefore, samples with a high percent of heavy elements cannot be analyzed with Compton Normalization. As a result, this calibration is used for very specific sample types.
  • FP calibration is an algorithmic approach based on the predictable detector response to known parameters, such as sample geometry, source energy, distance and angle between the detector and the sample, as well as other factors. FP calibration is preferred when a wide range of samples and matrices are analyzed. In such sample sets, FP can provide very precise data with good accuracy. Accuracy in the analysis can be increased by a cal-factor adjustment, which can be done internally in handheld analyzers and externally in FXL analyzers. The cal-factor adjustment uses a few known samples to adjust calibration by changing the slope and y-intercept of the factory calibration.
Read the application note, Factory-set FP Calibration Provides Precise Results in Well Site Geochemistry for data and analysis demonstrating the use of a portable XRF analyzer to deliver reliable, repeatable and accurate data on elements from Mg to U using FP factory calibration.
Share this article
8
SHARES
FacebookLinkedinTwitterMail
Petrochemical plant illuminated at twilight
Where Do Flowmeter Measurements Factor Into Downstream Oil and Gas?
In the downstream sector of the oil and gas industry—where... by Ethan Borchelt / 05.06.2025
Read More
Large industrial water treatment and boiler room. Shiny steel metal pipes and blue pumps and valves.
How to Better Detect Methane, CO, CO₂, and Volatile Organic Compounds (VOCs) Leaks
The oil and gas industry faces stringent regulatory complian... by Bob Gallagher / 02.18.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Trackbacks
  1. Technology from Space May Help Mining on Earth says:
    September 15, 2016 at 12:56 pm
    […] Portable XRF Calibrations in Well Site Geochemistry […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Ion Chromatography Simplifies Bromide Monitoring in Source Waters
New Dimensions at CIM Convention 2015
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook