  • Contact
Accelerating ScienceAdvancing Mining / Rare Earth Elements / Rare Earths Update
Rare Earths Update
Written by Marlene Gasdia-Cochrane | Published: 05.26.2016
bastnasite crystal
Bastnasite (crystal)
Several years ago we discussed how rare earth elements (REEs) are not very rare at all. Although originally thought to be rare, many of the minerals are actually common in the Earth’s crust.  However, they rarely exist in pure form and are usually found within other minerals, making them costly to mine.  (If you don’t know which elements make up the REEs, you can read this article and take a look at this infographic.)
Why are rare earth minerals important?  They play an important role in the increasing demand for clean and efficient energy.  Hybrid vehicles, permanent magnets, rechargeable batteries, wind turbines, compact fluorescent light bulbs, and catalytic converters — as well as cell phones, flat screen display panels, laptop computers, disk drives and more depend upon the unique properties of rare earth elements.
The U.S. Geological Survey, Mineral Commodity Summaries, January 2016 report was released and ironically there is currently an excess supply of rare earth elements.  The report reads in part:
“In 2015, excess supply caused prices for rare-earth compounds and metals to decline significantly, and China continued to dominate the global supply of rare earths. According to China’s Rare Earth Industry Association, consumption of rare-earth oxides in China was forecast to increase from 98,000 tons in 2015 to 149,000 tons in 2020. In 2015, China’s consumption was led by magnets (35%), abrasives (18%), and catalysts (15%). Illegal production was cited as a major factor in declining prices. According to sources in China, despite Government efforts, illegal production of rare-earth materials was ongoing. China’s rare-earths industry was in the process of being consolidated into six major industrial entities. The rare-earth mining production quota for 2015 was set at 105,000 tons, unchanged from 2014. China ended its rare-earth export quotas, removed export tariffs, and began to impose resource taxes on rare earths based on sales value instead of production quantity. Through October 2015, China had exported 26,800 tons of rare-earth materials, a 20% increase compared with exports for the same period in 2014. Production of rare-earth oxide equivalent in Malaysia, derived from Australian mine production, was 7,750 tons through September 2015, a 55% increase compared with the same period in 2014. U.S. domestic consumption of rare-earth compounds and metals was estimated to be nearly unchanged compared with that of 2014. In October, the Mountain Pass mining and separation operations were idled indefinitely. Price declines were cited as a key factor in the suspension of operations. The suspension resulted in a decline in mine production and exports of rare-earth compounds.”
Despite the excess supply, exploration has continued in the United States, as well as in several other countries — especially where bastnäsite and monazite deposits are found. Geochemical exploration is the main method of REE exploration. Portable XRF is a very useful tool to qualitatively and quantitatively evaluate REE projects in real time in the ­field. Portable XRF instruments can provide real-time, on-site assays of REEs and other elements in any type of geological samples, including elements associated with REE-bearing minerals. By using the concentrations from these elements, it is possible to infer concentrations of heavy REEs (HREEs) that are commonly associated with host minerals.
Ores of rare earth elements are mineralogically and chemically complex and commonly radioactive. Currently, REEs are extracted from the two mined minerals I mentioned: bastnasite and monazite.  However, sometimes rare earth elements are found where least expected.  Rare earth manganese nodules have been found beneath the Atlantic Ocean. A Popular Science article reported that “Last summer the UN’s International Seabed Authority issued the first deep sea exploration permits, allowing companies to start actively looking for places to mine nodules and other sources of rare earth elements from the ocean floor.”  Because of this find, a three-year research project involving 25 European research institutions will focus on analyzing the impact of deep sea mining on the seafloor environment.
What do you think about mining the seafloor to aid energy efficiency?   Comment below.
Share this article
FacebookLinkedinTwitterMail
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Comments
  1. Mehmet Salih Bayraktutan says
    June 4, 2016 at 7:16 am
    I found rich REE in geochem and isotopic analyses of my samples, from various arsas.turkey. during my post-doc
    Research study in Pisa italy geochemistry Ins- CNR…
    May l request your help in informing me about the economic limits of REE concentration, required for ree technology…
    Best Regards
    Reply
    • Marlene Gasdia-Cochrane says
      June 6, 2016 at 12:24 pm
      Mehmet Salih–Thank you for commenting. You can read additional article about rare earth elements in other blog articles:
      Click on the Rare Earth Elements section of the Advancing Mining blog:
      https://www.thermofisher.com/blog/mining/rare-earth-elements/
      Reply
Trackbacks
  1. From Tailings to Treasure? Miners Make Money Reprocessing Tailings says:
    June 14, 2016 at 1:34 pm
    […] elements; modern extraction techniques would now permit their recovery. This is good news because rare earth elements, which are extremely difficult and costly to mine, are critical components in consumer electronics […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Is There Enough Graphite to be Mined for the Electric Car Market?
Memorial Day 2016
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook