  • Contact
Accelerating ScienceAdvancing Mining / Cement / Ubiquitous Industrial Minerals: Nature’s Most Popular Raw Materials
Ubiquitous Industrial Minerals: Nature’s Most Popular Raw Materials
Written by Ali Somarin | Published: 06.03.2014
Industrial minerals are used, either in processed or natural state, to make building materials, paint, ceramics, glass, plastics, paper, electronics, detergents, medications and medical devices, and many more industrial and domestic products.
Industrial minerals are used, either in processed or natural state, to make building materials, paint, ceramics, glass, plastics, paper, electronics, detergents, medications and medical devices, and many more industrial and domestic products.
Do you know what industrial minerals are? You may not know them, but they permeate nearly every aspect of our daily lives. Industrial minerals are used, either in processed or natural state, to make building materials, paint, ceramics, glass, plastics, paper, electronics, detergents, medications and medical devices, and many more industrial and domestic products.
According to the Industrial Minerals Association of North America, every American consumes about 24 tons of industrial minerals each year.
Industrial minerals are generally defined as minerals that are not sources of metals, fuel, or gemstones. So what are they? The most widely-used industrial minerals include limestone, clays, sand, gravel, diatomite, kaolin, bentonite, silica, barite, gypsum, potash, pumice, and talc. Some of the industrial minerals commonly used in construction, such as crushed stone, sand, gravel, and cement, are called aggregates.
Industrial minerals are extremely versatile; most have at least two, sometimes many more, applications and span multiple markets. Talc, for example, is used in cosmetics, paper, and plastics. Silica sand is used to make glass, ceramics, and abrasives. While industrial minerals are defined as non-metallic, there are a few that have metallurgical properties, such as bauxite, which is the primary source of aluminum ore and is also used to make cement and abrasives. Bentonite and barite are non-fuel industrial minerals that have an important application in oil and gas extraction as components in drilling fluids. Bauxite and kaolin are used in fracking operations.
Industrial minerals are valued for their physical and chemical properties that make them so useful for so many products, and their price is driven by market demand for these items rather than by commodities exchange markets. Manufacturing, agriculture, and in particular the recovering construction and housing markets, are contributing to market growth for these minerals. For an in-depth look at the role of industrial minerals in the U.S economy, read the 2013 U.S. Geological Survey Mineral Commodity Summaries.
Market demand for industrial minerals also influences how they are mined. Industrial minerals are extracted primarily by surface mining, which is less expensive than underground mining. However, even when a location is determined to have a potentially economically viable mineral deposit, the costs of drilling, extraction, and transporting the raw materials still must be considered and weighed against the current market demand for that particular mineral. Industrial minerals are typically mined from existing sites or areas that are close to infrastructure as their price usually doesn’t justify the cost of building up the infrastructure needed to explore a new site.
Before a mining plan is developed, geologists need to map out the mineral distribution of the deposit by evaluating the geological processes, also called mineralizing events, which formed them. Once it’s been determined that a sufficient quantity of minerals exists and cost-effective mining can begin, geologists study the lithology and other geochemical data to direct and control the mining process.  This is where X-ray fluorescence (XRF) analysis can assist. XRF is one of the most advanced tools for exploration and mining of industrial minerals. Portable XRF analyzers are an emerging instrument of choice for in-quarry exploration and evaluating the composition of raw materials such as phosphate, potash, gypsum, and limestone for industrial use. Other useful applications for portable XRF analysis in industrial minerals mining include:
  • Determining penalty elements in limestone, Fe ore, and bauxite
  • Blending and sorting of raw materials
  • Flagging grade, sub-grade, and waste, and prevent taking ore to the waste heap.
Lab-based XRF is a complimentary technique for evaluating prepared mineral samples for quality control and to determine their suitability for specific applications. Please share your experiences with either a portable or lab-based XRF analyzer for industrial minerals applications. As always, we welcome your comments.
Share this article
231
SHARES
FacebookLinkedinTwitterMail
Engineers and staff communicate on site in chemical plant
Why Flow Computing is Key to Minimizing Disruptions in Station Control
Today’s station control systems—whether in oil and gas, ... by Ethan Borchelt / 04.08.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Comments
  1. Summer Forlonge says
    January 18, 2021 at 10:08 am
    Very thankful for this wonderful article, it seem like it will really help with what I need to get done. Thank you very much for the post!
    Reply
  2. Anand Talc says
    July 22, 2021 at 4:41 am
    thank you for sharing valueable post
    Reply
  3. Himanshu Khatwanii says
    August 2, 2021 at 9:33 am
    great!!
    thank you for sharing post from here you
    Reply
  4. Rajveer Singh Rathore says
    March 26, 2022 at 10:25 am
    Thank you for sharing this valuable post this post is very informative and useful to me.
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Fascinating Facts About Manganese
Euro Mine Expo in Swedish Lapland
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook