693f - Key/Specs/Resources/Samples/Tech/Doc/Contact
X-ray photoelectron spectroscopy
The Thermo Scientific Nexsa G2 X-Ray Photoelectron Spectrometer (XPS) System offers fully automated, high-throughput surface analysis, delivering the data to advance research and development or to solve production problems. The integration of XPS with ion scattering spectroscopy (ISS), UV photoelectron spectroscopy (UPS), reflected electron energy loss spectroscopy (REELS), and Raman spectroscopy, allows you to conduct true correlative analysis. The system now includes options for sample heating and sample biasing capabilities to increase the range of experiments now possible. The Nexsa G2 Surface Analysis System unlocks the potential for advances in materials science, microelectronics, nanotechnology development, and many other applications.
Video Player is loading.
Current Time 0:00
Duration 4:12
Loaded: 3.89%
Stream Type LIVE
Remaining Time 4:12
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    The Nexsa G2 System: An Introduction with Tim Nunney
    Key Features
    High-performance X-ray source
    High-performance X-ray source
    A new, low-power X-ray monochromator allows selection of the analysis area from 10 µm to 400 µm in 5 µm steps, ensuring data is collected from the feature of interest while maximizing the signal.
    Optimized electron optics
    Optimized electron optics
    The high-efficiency electron lens, hemispherical analyzer, and detector allow for superb detectability and rapid data acquisition.
    Sample viewing
    Sample viewing
    Bring sample features into focus with the Nexsa XPS System's patented optical viewing system and XPS SnapMap, which helps you pinpoint areas of interest quickly using a fully focused XPS image. 
    Insulator analysis
    Insulator analysis
    The patented dual-beam flood source couples low-energy ion beams with very low-energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need for charge referencing, making the analysis of the data from insulating samples easy and reliable. 
    Depth profiling
    Depth profiling
    Go beyond the surface with a standard ion source or MAGCIS, the optional dual-mode monatomic and gas cluster ion source; automated source optimization and gas handling ensure excellent performance and experimental reproducibility.
    Optional sample holders
    Optional sample holders
    Specialist sample holders for angle-resolved XPS, sample bias measurements, or for inert transfer from a glove box are available.
    Digital Control
    Digital Control
    Instrument control, data processing, and reporting are all controlled from the Windows Software-based Avantage data system. 
    NX sample heater module
    Fully software-controlled sample heating option, enabling temperature-dependent studies.
    Video Player is loading.
    Current Time 0:00
    Duration 1:33
    Loaded: 10.61%
    Stream Type LIVE
    Remaining Time 1:33
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Animation: Thermo Scientific Nexsa surface analysis system
      Specifications
      Style Sheet for Products Table Specifications
      Analyzer type
      • 180°, double-focusing, hemispherical analyzer with 128-channel detector
      X-ray source type
      • Monochromated, micro-focused, low-power Al K-Alpha X-ray source
      X-ray spot size
      • 10–400 µm (adjustable in 5 µm steps)
      Depth profiling
      • EX06 monatomic ion source or MAGCIS dual-mode ion source
      Maximum Sample area
      • 60 x 60 mm
      Maximum sample thickness
      • 20 mm 
      Vacuum system
      • Two turbo molecular pumps, with automated titanium sublimation pump and backing pump 
      Optional accessories
      • UPS, ISS, REELS, iXR Raman spectrometer, MAGCIS, sample tilt module, NX sample heating module, sample bias module, vacuum transfer module, adaptor for glove box integration 
      Style Sheet for Komodo Tabs
      Resources
      Thermo Scientific Nexsa Surface Analysis System
      High sensitivity for large areas, small X-ray spots for features and integrated surface analysis techniques, the Nexsa system has it all!
      Video Player is loading.
      Current Time 0:00
      Duration 3:16
      Loaded: 5.04%
      Stream Type LIVE
      Remaining Time 3:16
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
          1. Now Playing
            Up NextThermo Scientific Nexsa Surface Analysis System
            High sensitivity for large areas, small X-ray spots for features and integrated surface analysis techniques, the Nexsa system has it all!
          2. Now Playing
            Up NextWebinar: Thermo Scientific Nexsa Surface Analysis System
            The Nexsa System is a high-performance X-ray Photoelectron Spectrometer, designed to integrate other analytical techniques while maintaining a high-throughput workflow.
          3. Now Playing
            Up NextPart 1- Understanding Surface Chemistry with Avantage Software
            Understanding Surface Chemistry with Avantage Software
          4. Now Playing
            Up NextPart 2 - Understanding XPS images and depth profiles with Avantage Software
            By watching the webinar, you will learn how to: Use the right tools to understand multi-level data sets, such as depth profiles and images. Apply multi-variate techniques (such as PCA) to understand XPS images. Peak fit depth profile data sets to
          5. Now Playing
            Up NextThe Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
            This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
          6. Now Playing
            Up NextMulti-technique and Complementary Analysis session (part 2) of the Ignite Surface Analysis Virtual Scientific Conference
            The speakers Roland Barbosa (Covalent Metrology), Paul Mack and Herman Lemmens (Thermo Fisher Scientific) give presentations on multi-technique and complementary analysis.
        Watch on-demand: Thermo Scientific Nexsa G2 Surface Analysis System Demo
        You will learn how the Nexsa G2 XPS System can be used to investigate a wide range of materials using XPS, supported by the additional analysis techniques on the system: UV photoelectron spectroscopy (UPS), ion scattering spectroscopy (ISS), reflected electron energy loss spectroscopy (REELS), Raman spectroscopy, and more.
        Register to watch
        Thermo Scientific Nexsa Surface Analysis System
        High sensitivity for large areas, small X-ray spots for features and integrated surface analysis techniques, the Nexsa system has it all!
        Video Player is loading.
        Current Time 0:00
        Duration 3:16
        Loaded: 5.04%
        Stream Type LIVE
        Remaining Time 3:16
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
            1. Now Playing
              Up NextThermo Scientific Nexsa Surface Analysis System
              High sensitivity for large areas, small X-ray spots for features and integrated surface analysis techniques, the Nexsa system has it all!
            2. Now Playing
              Up NextWebinar: Thermo Scientific Nexsa Surface Analysis System
              The Nexsa System is a high-performance X-ray Photoelectron Spectrometer, designed to integrate other analytical techniques while maintaining a high-throughput workflow.
            3. Now Playing
              Up NextPart 1- Understanding Surface Chemistry with Avantage Software
              Understanding Surface Chemistry with Avantage Software
            4. Now Playing
              Up NextPart 2 - Understanding XPS images and depth profiles with Avantage Software
              By watching the webinar, you will learn how to: Use the right tools to understand multi-level data sets, such as depth profiles and images. Apply multi-variate techniques (such as PCA) to understand XPS images. Peak fit depth profile data sets to
            5. Now Playing
              Up NextThe Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
              This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
            6. Now Playing
              Up NextMulti-technique and Complementary Analysis session (part 2) of the Ignite Surface Analysis Virtual Scientific Conference
              The speakers Roland Barbosa (Covalent Metrology), Paul Mack and Herman Lemmens (Thermo Fisher Scientific) give presentations on multi-technique and complementary analysis.
          Watch on-demand: Thermo Scientific Nexsa G2 Surface Analysis System Demo
          You will learn how the Nexsa G2 XPS System can be used to investigate a wide range of materials using XPS, supported by the additional analysis techniques on the system: UV photoelectron spectroscopy (UPS), ion scattering spectroscopy (ISS), reflected electron energy loss spectroscopy (REELS), Raman spectroscopy, and more.
          Register to watch
          Style Sheet to change H3 to p with em-h3-header class
          Style Sheet to change Sample H3 to p with em-h3-header class
          Samples
          Battery Research
          Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
          Learn more ›
          Metals Research
          Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
          Learn more ›
          Polymers Research
          Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
          Learn more ›
          Geological Research
          Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
          Learn more ›
          Oil and Gas
          As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
          Learn more ›
          Nanoparticles
          Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
          Learn more ›
          Forensics
          Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
          Learn more ›
          Catalysis Research
          Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
          Learn more ›
          Fibers and Filters
          The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
          Learn more ›
          2D Materials
          Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
          Learn more ›
          Automotive Materials Testing
          Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
          Learn more ›
          Techniques
          Multi-technique surface analysis workflow
          To meet the need for extensive characterization of surfaces, we have established multi-technique workflows based on using either the Thermo Scientific ESCALAB CXi XPS Microprobe or the Thermo Scientific Nexsa Surface Analysis System. These instruments are designed as multi-technique workstations to provide comprehensive analyses in a timely and efficient manner. 
          Learn more ›
          X-Ray Photoelectron Spectroscopy
          X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
          Learn more ›
          Multi-technique surface analysis workflow
          To meet the need for extensive characterization of surfaces, we have established multi-technique workflows based on using either the Thermo Scientific ESCALAB CXi XPS Microprobe or the Thermo Scientific Nexsa Surface Analysis System. These instruments are designed as multi-technique workstations to provide comprehensive analyses in a timely and efficient manner. 
          Learn more ›
          X-Ray Photoelectron Spectroscopy
          X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
          Learn more ›
          Style Sheet to change H2 style to p with em-h2-header class
          Style Sheet to change H2 style to p with em-h2-header class
          Contact us
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards