Plants showcase
Genomics tools for improved plant breeding and production
The discovery of novel single nucleotide polymorphisms (SNPs) is critical for plant genotyping in that it helps to provide candidate markers for marker-assisted selection programs in key crops. Genomic approaches are useful for accelerating variety development and crop improvement. Genomic tools can assist in the detection of QTLs, favorable and deleterious traits and identification of favorable alleles. Thermo Fisher Scientific offers a range of genomic tools and resources to help facilitate your plant breeding and production goals.
Quick links
AgriSeq targeted genotyping-by-sequencing (GBS)
Applied Biosystems AgriSeq targeted genotyping-by-sequencing (GBS) solutions allow for high-throughput analysis of plant genotypes in a flexible and cost-effective manner. Leveraging a highly efficient multiplex chemistry, hundreds to thousands of genetic loci can be simultaneously targeted and efficiently amplified in a single reaction. Libraries are then processed for next-generation sequencing (NGS) where hundreds of samples can be barcoded and sequenced in the same run. Capable of generating up to 2.6 million genotypes per day from high-quality NGS data at pennies per data point, AgriSeq targeted GBS represents the future of technology to help advance your plant research.
AgriSeq chemistry is designed for analysis of SNP panels of 50–5,000 markers. Customized panels designed to target your relevant markers will help to accelerate variety development and crop improvement. Our dedicated team of agrigenomics bioinformatics professionals have designed a range of successful panels for plants ranging from corn, soybean, tomato, cotton, cucumber, spinach, and many more.
Axiom microarrays
Our array-based genotyping solutions are ideal for the identification, validation and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays or customize an array specifically for your study.
Product
No. of markers
Array Format
Cat. No.
Price
Axiom Citrus HD Genotyping Array (Citrus15 AX1)
702,244
96-array
550729
Contact a sales representative ›
Axiom Citrus HD Genotyping Array (Citrus15 AX2)
702,198
96-array
550730
Contact a sales representative ›
Axiom Citrus Genotyping Array
58,433
384-array
550775
Contact a sales representative ›
Axiom Peanut Genotyping Array
47,837
384-array
550846
Contact a sales representative ›
Axiom Peanut Genotyping Array
47,837
Mini 96-array
550865
Contact a sales representative ›
Axiom Rose Genotyping Array
137,786
96-array
550450
Contact a sales representative ›
Axiom Rye Genotyping Array
600,843
96-array
550535
Contact a sales representative ›
Axiom Strawberry Genotyping Array (i35)
34,260
384-array
550767
Contact a sales representative ›
Axiom Strawberry Genotyping Array (i35)
34,260
Mini 96-array
550827
Contact a sales representative ›
Axiom Tomato Genotyping Array
51,214
96-array
550497
Contact a sales representative ›
Custom TaqMan SNP Genotyping Assays
These assays provide a highly flexible technology for detection of polymorphisms within any genome. Create your own assays by submitting target sequences to our secure assay design pipeline using the Applied Biosystems Custom TaqMan Assay Design Tool. This pipeline has successfully generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. TaqMan Assays can be run on any of our real-time PCR instruments or used as an endpoint assay on other third-party systems, including the Array Tape platform.
Recommended reagents
Catalog # Name Size Price (USD) Qty
A30872 TaqPath™ ProAmp™ Master Mix, 2 x 50 mL Each
8,602.65
Online Exclusive
9,280.00
Save 677.35 (7%)
A30867 TaqPath™ ProAmp™ Master Mix, 1 x 50 mL Each
4,380.65
Online Exclusive
4,725.00
Save 344.35 (7%)
A30871 TaqPath™ ProAmp™ Master Mix, 2 x 10 mL Each
1,888.65
Online Exclusive
2,035.00
Save 146.35 (7%)
A30866 TaqPath™ ProAmp™ Master Mix, 1 x 10 mL Each
1,019.65
Online Exclusive
1,100.00
Save 80.35 (7%)
A30865 TaqPath™ ProAmp™ Master Mix, 1 x 1 mL Each
123.65
Online Exclusive
134.00
Save 10.35 (8%)
For Research Use Only. Not for use in diagnostic procedures.