Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 30.0 | 0.4 |
L-Alanine | 89.0 | 25.0 | 0.28089887 |
L-Arginine hydrochloride | 211.0 | 84.0 | 0.39810428 |
L-Asparagine-H2O | 150.0 | 28.4 | 0.18933333 |
L-Aspartic acid | 133.0 | 30.0 | 0.22556391 |
L-Cystine | 240.0 | 70.0 | 0.29166666 |
L-Glutamic Acid | 147.0 | 75.0 | 0.5102041 |
L-Glutamine | 146.0 | 584.0 | 4.0 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 105.0 | 0.8015267 |
L-Leucine | 131.0 | 105.0 | 0.8015267 |
L-Lysine hydrochloride | 183.0 | 146.0 | 0.7978142 |
L-Methionine | 149.0 | 30.0 | 0.20134228 |
L-Phenylalanine | 165.0 | 66.0 | 0.4 |
L-Proline | 115.0 | 40.0 | 0.3478261 |
L-Serine | 105.0 | 42.0 | 0.4 |
L-Threonine | 119.0 | 95.0 | 0.79831934 |
L-Tryptophan | 204.0 | 16.0 | 0.078431375 |
L-Tyrosine disodium salt | 225.0 | 104.0 | 0.46222222 |
L-Valine | 117.0 | 94.0 | 0.8034188 |
Vitamins | |||
Biotin | 244.0 | 0.013 | 5.327869E-5 |
Choline chloride | 140.0 | 4.0 | 0.028571429 |
D-Calcium pantothenate | 477.0 | 4.0 | 0.008385744 |
Folic Acid | 441.0 | 4.0 | 0.009070295 |
Niacinamide | 122.0 | 4.0 | 0.032786883 |
Pyridoxal hydrochloride | 204.0 | 4.0 | 0.019607844 |
Riboflavin | 376.0 | 0.4 | 0.0010638298 |
Thiamine hydrochloride | 337.0 | 4.0 | 0.011869436 |
Vitamin B12 | 1355.0 | 0.013 | 9.594096E-6 |
i-Inositol | 180.0 | 7.2 | 0.04 |
Inorganic Salts | |||
Calcium Chloride (CaCl2-2H2O) | 147.0 | 219.0 | 1.4897959 |
Magnesium Sulfate (MgSO4-2H2O | 176.0 | 200.0 | 1.1363636 |
Potassium Chloride (KCl) | 75.0 | 330.0 | 4.4 |
Potassium Nitrate (KNO3) | 101.0 | 0.076 | 7.524752E-4 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 3024.0 | 36.0 |
Sodium Chloride (NaCl) | 58.0 | 4505.0 | 77.67242 |
Sodium Phosphate monobasic (NaH2PO4-2H2O) | 156.0 | 141.0 | 0.90384614 |
Sodium Selenite (Na2SeO3-5H20) | 263.0 | 0.0173 | 6.5779466E-5 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 4500.0 | 25.0 |
HEPES | 238.0 | 5958.0 | 25.033613 |
Phenol Red | 376.4 | 15.0 | 0.039851222 |
Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
1. Dulbecco, R. and Freeman, G. (1959) Virology 8:396.
2. Iscove, N.N. and Melchers, F. (1978) J. Exper. Med., 147:923.
3. Smith, J.D., Freeman, G., Vogt, M., et al., (1960) Virology 12:185.
12440 - IMDM
Cat No:
12440046
,
12440053
,
12440061
,
12440079