Cultured axolotl sensory neurons. Anti–human neuronal protein HuC/HuD antibody, Alexa Fluor® 546 goat anti–mouse IgG antibody, Alexa Fluor® 488 phalloidin and Hoechst 33342.
Cultured axolotl sensory neurons stained with our anti–human neuronal protein HuC/HuD antibody (anti-Hu, Cat. no. A21271) and detected using the red-orange–fluorescent Alexa Fluor® 546 goat anti–mouse IgG antibody (Cat. no. A11003). Actin was labeled using green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379) and nuclei were stained with blue-fluorescent Hoechst 33342 (Cat. no. H1399, H3570, H21492). The image was contributed by Josh Gross and Linda Barlow, University of Denver, Denver, Colorado.
Cultured axolotl sensory neurons stained with our anti–human neuronal protein HuC/HuD antibody (anti-Hu, Cat. no. A21271) and detected using the red-orange–fluorescent Alexa Fluor® 546 goat anti–mouse IgG antibody (Cat. no. A11003).  Actin was labeled using green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379) and nuclei were stained with blue-fluorescent Hoechst 33342 (Cat. no. H1399, H3570, H21492).  The image was contributed by Josh Gross and Linda Barlow, University of Denver, Denver, Colorado.
Related Products
Catalog # Name Size Price (USD) Qty
A11003 Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 546 1 mg
290.00
A12379 Alexa Fluor™ 488 Phalloidin, Green, 495/518 nm Each
644.65
Online Exclusive
664.00
Save 19.35 (3%)
A21271 HuC/HuD Monoclonal Antibody (16A11) 100 µg
488.00
H1399 Hoechst 33342, trihydrochloride trihydrate, 100 mg, 100 mg Each
109.65
Online Exclusive
113.00
Save 3.35 (3%)
Related Images