Antibody (702241) in Flow
Flow Cytometry analysis of endogenous 14-3-3 protein epsilon / YWHAE was performed on HeLa cells labeled with ABfinity? Anti-14-3-3 protein epsilon / YWHAE Recombinant Rabbit Monoclonal Antibody (Product ## 702241, 5 ug/ 1M cells) or with rabbit isotype control and detected with Goat anti-Rabbit IgG (H+L) Superclonal? Secondary Antibody, (Alexa Fluor® 488 conjugate, Product ## A27034, 0.4 ug/ml, 1:2500) as represented by the red and pink histograms respectively. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control. A representative of 10,000 cells were acquired and analyzed for each sample using an Attune® Acoustic Focusing Cytometer (4468770).
Flow Cytometry analysis of endogenous 14-3-3 protein epsilon / YWHAE was performed on HeLa cells labeled with ABfinity? Anti-14-3-3 protein epsilon / YWHAE Recombinant Rabbit Monoclonal Antibody (Product ## 702241, 5 ug/ 1M cells) or with rabbit isotype control and detected with Goat anti-Rabbit IgG (H+L) Superclonal? Secondary Antibody, (Alexa Fluor® 488 conjugate, Product ## A27034, 0.4 ug/ml, 1:2500) as represented by the red and pink histograms respectively. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control. A representative of 10,000 cells were acquired and analyzed for each sample using an Attune® Acoustic Focusing Cytometer (4468770).
Related Products
Catalog # Name Size Price (USD) Qty
702241 14-3-3 epsilon Recombinant Rabbit Monoclonal Antibody (5H10L5) 100 µg
504.00
Related Images