Overview
The Wolff-Kishner Reduction is a prominent process for the deoxygenation of aldehydes and ketones to their corresponding hydrocarbons
Wolff-Kishner Reduction overview
Named Reactions in Organic Synthesis
Wolff-Kishner Reduction Mechanism
Watch our video to learn more about the Wolff-Kishner Reduction and its mechanisms. 
Video Player is loading.
Current Time 0:00
Duration 2:16
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Wolff-Kishner Reduction?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Wolff-Kishner Reduction. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Wolff-Kishner Reduction
    In 1911, Nikolai Kishner added a hydrazone dropwise to a mixture of hot potassium hydroxide and a platinized porous plate, and the corresponding hydrocarbon was formed. A year later, Ludwig Wolff demonstrated that heating an ethanolic solution of semicarbazones and hydrazones in a sealed tube at approximately 180°C in the presence of sodium ethoxide gave the same result. The deoxygenation of aldehydes and ketones to their corresponding hydrocarbons is now called the Wolff-Kishner reduction.
    Since the original experiments, the procedure has undergone substantial modifications to allow the use of milder reaction conditions and to expand the number of substrates, as well as to increase yields. For many years, the standard methodology involved mixing the carbonyl compound with neat hydrazine in a high-boiling solvent such as ethylene glycol in the presence of excess base such as sodium ethoxide. However, the reaction often required refluxing for several days because of the temperature-lowering effects of the water produced as a by-product of hydrazine formation.
    The Huang-Minion modification involved removing the water and excess hydrazine via distillation, allowing the reaction temperature to increase once the hydrazine formation was complete. This dramatically shortened the reaction time to just a few hours and allowed the use of the less expensive reagent hydrazine hydrate, along with water-soluble bases such as sodium hydroxide.
    In another modified procedure, known as the Caglioti reaction, tosylhydrazones are used with hydride reagents to obtain the corresponding alkynes.
    The total synthesis of dysidiolide, the first compound found to be a natural inhibitor of protein phosphatase cdc25A, which is essential for cell proliferation, utilized the Wolff-Kishner reduction in the production of an advanced bicyclic intermediate.
    Other Reduction Reactions
    Other Reduction Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:00