Why use array tomography for volume electron microscopy?
Array tomography can provide rapid localization of cells and their interaction partners in tissues.  With the largest imaging field, array tomography is ideal for tissue histology.  Ultrastructure analysis of whole animals or whole organs is not needed, so array tomography provides easy navigation for samples that can be stored and re-imaged.
Turn any Thermo Scientific SEM into a volume EM microscope with the addition of automated Array tomography.
Mouse brain sample recorded on an Apreo 2 SEM using array tomography. Array tomography analysis of a mouse brain
How does array tomography SEM work?
Thermo Scientific Maps Correlative Electron Microscopy Software guides the acquisition of 3D volumes for array tomography. When a user employs an SEM-based method for 3D reconstruction, the total acquisition time can quickly become impractical if the selected volume is unnecessarily large. In that case, the ability to precisely identify an area of interest becomes particularly important. Given the capacity to create large overviews at high resolution and the correlation with fluorescent images, Maps provides the 2D map to target 3D acquisitions (e.g., using Thermo Scientific Auto Slice & View Software), thus delivering results more quickly. 
Additionally, the modular architecture of Maps software allows integration of more complex, SEM-based, 3D acquisition schemes. Maps software allows users to define multiple imaging areas on the block face and guides the user through the setup of slicing and imaging parameters. With the optional array tomography plugin for SEM/DualBeam systems, Maps software assists the user in imaging serial sections for 3D reconstruction.
Graphical depiction of array tomography
Amira Software rendering of the drosophila gut
Thermo Scientific Amira software volume rendering of a very large 3D data set recorded in 80 sections at 3.9 nm resolution on a Helios G4 Plasma-FIB-SEM. 
Amira Software volme rendering of array tomography data from a drosophila gut
Meet the scientists using array tomography
Narayanan Kasthuri from University of Chicago
“In Maps with array tomography, less than 10µm section-to-section variability of imaging region placement allows imaging with much less overhead. This variability can be hundreds of microns in other solutions. Imaging with less overhead speeds up time to data by a large factor especially when imaging small target structures.”
Narayanan 'Bobby' Kasthuri, Neuroscientist, University of Chicago
 Irnia Kolotuev, EM facility coordinator from University of Lausanne.
“Electron microscopy analysis is frequently complex. Our array tomography protocol with Maps provides a more straightforward electron microscopy data analysis. This facilitates processing and acquisition of the EM data from the large sample surface and intermediate volume. Given the overall ease of the approach, we believe that it can be used to address not only scientific questions, but also as a diagnostic tool.” 
Irnia Kolotuev, EM Facility Coordinator, University of Lausanne
Array tomography resources
Maps Software for Array Tomography
Thermo Scientific Maps Software is an imaging and correlative workflow software suite compatible with the full line of Thermo Scientific SEM, FIB SEM and TEM platforms.
Automated acquisition of large microscopy images at high resolution
Protocol: Targeted 3D EM and Correlative Microscopy Method using SEM Array Tomography
Optimized scanning electron microscopy workflow for volume array tomography for asymmetric samples and model organisms. 
Array tomography of nonlinear sample, C. elegans larva.