Find the best fit for your business
X-ray has grown to become the most popular detection technology in food processing because it offers expanded inspection capabilities over just metal detection. We offer flexible solutions that span from easy to use, affordable workhorse models to highly specialized, powerful systems to meet particular application requirements.  View the comparison table below to better understand the most common selection criteria to determine which system best meets your needs, or use the Contact Sales button above to get in touch with a specialist for assistance.
Compare food x-ray systems
NextGuard C330
NextGuard C500
NextGuard Pro
Xpert Conveyor
Xpert Bulk
Xpert Sideshoot
Max Inspectable Area
13.4 in. wi. x
7.3 in. ht.
19.7 in. wi. x
9.8 in. ht.
19.7 in. wi. x
9.8 in. ht.
15.7 in. wi. x
10 in. ht.
19.4 in. wi. x
2 in. ht.
7 in. wi. x
10.8 in. ht.
Package Types Supported
Cartons/Boxes
Pouches/Bags
Bulk Flow Products
Plastic Bottles and Cans
Washdown Rating
IP65 with AC
IP65
IP65, stainless steel bearings and gear motor
IP65 standard, IP69K optional
IP65
IP65
Typical Sensitivity
0.8–1.5 mm SST, 2–5 mm glass
1.5–2 mm SST, 4–7 mm glass
Same as standard models
1–2 mm SST, 4–7 mm glass
1–2 mm SST, 3–5 mm glass
1–2 mm SST, 3–5 mm glass
Max X-Ray Power
160W
200W
Same as standard models
315W
315W
315W
Relative Price
$
$$
$$$
$$$$
$$$$
$$$$
Max Product Weight
13 lbs
24 lbs
110 lbs
88 lbs
88 lbs
N/A external conveyor
X-Ray Beam Orientation
Vertical, bottom up, no blind spots
Vertical, bottom up, no blind spots
Vertical, bottom up, no blind spots
Vertical, top down
Vertical, top down
Horizontal, back to front
Belt Speed Range
10-120 m/min
5-50 m/min
5-120 m/min
5-60 m/min
5-60 m/min
5-80 m/min
Single Conveyor with Reject Integrated
N/A external conveyor
Multi-Lane Operation
Remote Technical Support
1x1 image pixel for data collection