Thermo Scientific FTIR spectrometer and microscope resources
Learn more about FTIR spectroscopy and get the resources you need
Here you can find everything FTIR, from the most basic (what do “FT” and “IR” mean?) to applications using hyphenated techniques, like TGA-IR. Along the way, you can find information on sample preparation for transmission experiments, the power of FTIR Microscopy and charts relating chemical structure to the infrared spectrum. Come back often, as this content is being updated continually.
FTIR basics
Your starting point for learning about FTIR, this section covers what “FTIR” stands for and how it works. We also introduce the main sampling techniques and provide a more extensive overview of how they can be used to answer your questions.
Introduction to FTIR spectroscopy
FTIR spectroscopy offers a vast array of analytical opportunities in academic, analytical, QA/QC and forensic labs. Deeply ingrained in everything from simple compound identification to process and regulatory monitoring, FTIR covers a wide range of chemical applications, especially for polymers and organic compounds. Learn more about the basics and the value of this popular technique by watching the tutorials below. Videos also provide an overview of two common detectors and the apodization function.
What is FTIR?
FTIR stands for Fourier transform infrared, the preferred method of infrared spectroscopy. When IR radiation is passed through a sample, some radiation is absorbed by the sample and some passes through (is transmitted). The resulting signal at the detector is a spectrum representing a molecular ‘fingerprint’ of the sample. The usefulness of infrared spectroscopy arises because different chemical structures (molecules) produce different spectral fingerprints.
So, what is FTIR?
The Fourier Transform converts the detector output to an interpretable spectrum.
The FTIR generates spectra with patterns that provide structural insights.
Watch the tutorial for a quick description of what "FTIR" means and how the "FT" and the "IR" parts work together.
What is FTIR
How does FTIR work and why use it?
The FTIR uses interferometry to record information about a material placed in the IR beam. The Fourier Transform results in spectra that analysts can use to identify or quantify the material.
  • An FTIR spectrum arises from interferograms being ‘decoded’ into recognizable spectra
  • Patterns in spectra help identify the sample, since molecules exhibit specific IR fingerprints
How does FTIR work and why use it?
FTIR sampling introduction
There are four major sampling techniques in FTIR:
  • Transmission
  • Attenuated Total Reflection (ATR)
  • Specular Reflection
  • Diffuse Reflectance
Each technique has strengths and weaknesses which motivate their use for specific samples
FTIR sampling introduction
Sampling and applications of FTIR
FTIR can be a single purpose tool or a highly flexible research instrument. With the FTIR configured to use a specific sampling device – transmission or ATR for instance – the spectrometer can provide a wide range of information:
  • Most commonly, the identification of an unknown
  • Quantitative information, such as additives or contaminants
  • Kinetic information through the growth or decay of infrared absorptions
  • Or more complex information when coupled with other devices such as TGA, GC or Rheometer
Ultimately, FTIR can be a cost-effective answer machine.
Watch the tutorial for a more extensive examination of FTIR sampling techniques, including hyphenated sampling. Examples are shown and discussed giving an overview of what is possible.
Sampling and applications of FTIR
Featured FTIR detector videos
Featured FTIR apodization videos
Reference charts for spectroscopy and spectrometry
Organic Functional Chart
Functional groups are structural units within organic compounds defined by specific atom and bond arrangements. Infrared is a powerful identification tool for functional groups because of the similar absorption frequencies for those groups in different molecules. The actual frequency is affected by the environment, so the reference chart shows wide bands rather than specific frequencies. The identification of functional groups is a cornerstone of IR spectroscopy and organic chemistry.
Organic Functional Chart
Spectral Range Chart
Flexible FTIR spectrometers, such as the Thermo Scientific Nicolet line of FTIR spectrometers, can be configured to cover a wide range of performance. A part of this is spectral range as shown in this chart, where certain combinations of components provide high performance in specific ranges. There are often trade-offs, such as between high sensitivity using an MCT-A liquid-nitrogen cooled detector versus the wider spectral range but lower sensitivity of the DLaTGS room temperature detector.
Spectral Range Chart
FTIR sampling techniques
An in-depth training section, this will cover transmission, reflection, DRIFTS and ATR accessories, their use and care. The reasons for choosing one or the other method will become clear, and the strengths of each method will be highlighted. Stress here is on sample preparation and experiment optimization.
Why is it important to know about different methods of sample handling?
Certain techniques of sample handling are more effective than others for specific sample types. To obtain the best quality spectrum from your sample, it is important to know which handling technique works best for your sample type. Acquiring the best spectral data possible will give you more confidence in your results.
FTIR sample techniques
Explore these four sample handling techniques to learn about how they work, what types of samples you can analyze and the advantages to using each technique.
FAQs
Our market leading expertise in FTIR and FTIR microscopy is not just about the instruments, but it is due to our wide experience and knowledgeable staff. We have gathered here many of the questions we have been asked and will add to this section frequently.