QuantiGene Plex Data Analysis Software Icon
Streamlined gene expression analysis
Analyze data generated from QuantiGene Plex (QGP) assays with two freely available software options: QuantiGene Plex Data Analysis Software and Applied Biosystems Transcriptome Analysis Console (TAC) Software. These powerful software applications provide streamlined analysis and advanced visualization so you can get the most out of your data.
Data analysis workflow
Data analysis workflow for QuantiGene Plex Gene Expression Assays
QuantiGene Plex Data Analysis Software (Thermo Fisher Connect) introduces an improved workflow that provides customization capabilities for normalization and analysis of gene expression data generated using the QuantiGene Plex assay. Data from QuantiGene Plex Data Analysis Software can be exported into Transcriptome Analysis Console (TAC) Software for advanced analysis.
QuantiGene Plex Data Analysis Software
QuantiGene Plex Data Analysis Software is free software available through Connect, the Thermo Fisher Scientific cloud-based platform, that provides easy-to-follow workflows to improve data analysis of QuantiGene Plex assays. Data collected from QuantiGene Plex assays can be exported to Connect and conveniently accessed via web whenever an internet connection is available. Each Connect account offers 1 TB of cloud storage at no charge.
Access software
Software capabilities:
  • Control and treatment group assignments
  • Customize QC parameters
  • Analyze data from both 96-well and 384-well plates
  • Perform linearity of dilution calculations
  • Calculation of p-values during fold change analysis
  • Rank normalization genes for your analysis
  • View and export mean and individual sample values
  • Group samples and analyze the fold change in up to five groups
  • Remove samples or targets that do not meet QC criteria
  • Proceed directly to analysis without assigning background wells
Software workspaceFunctionFeatures
Setup plate map
Plate map set up
Set up plate for analysisAssign:
  • Samples to different groups for comparison
  • Background wells 
  • Wells that should not be included in the analysis
Normalization
Normalize data
Select and edit normalization genes and calculate assay linearity
  • Normalization ranking to calculate the stability measure (M) for each selected gene
  • Calculate assay linearity and % recovery
Review and edit QC criteria
Review and edit QC criteria
View and adjust the thresholds for QC parameters for data analysis
  • Maximum background
  • Limit of Detection
  • Technical Precision (%CV)
  • Saturation
  • Minimum Bead Count
Run analysis
  Run analysis
Run and analyze normalized results of differential gene expression analysisCalculate/view:
  • Change in expression of selected targets with respect to the reference sample or reference group and view graph
Create an account today
Already have an account? Access the QuantiGene Plex application.
Learn more about Thermo Fisher Cloud data security
Software highlights
Transcriptome Analysis Console
For enhanced data visualization and analysis beyond simple identification of differential expression, data can be exported from the QuantiGene Plex Data Analysis Software platform into Applied Biosystems Transcriptome Analysis Console (TAC) Software. Find tools to create scatter and volcano plots and perform hierarchical clustering. In addition, link out to publicly available annotations.
TAC Software capabilities:
  • Perform QC and data normalization
  • Perform statistical tests for differential expression
  • Focus on genes or pathways of interest
  • Explore interactions between coding and non-coding RNA
  • Link out to publicly available annotations
  • Obtain sequence information to design validation experiments
Download the latest version of TAC Software (64-bit). This version is supported on Microsoft Windows™ 7 (SP 1) and Windows 10 Professional.
Download  Release notes
Video: How to set up an Invitrogen™ QuantiGene™ Plex Assay in the Luminex® xPONENT® Software
Video Player is loading.
Current Time 0:00
Duration 8:11
Loaded: 0%
Stream Type LIVE
Remaining Time 8:11
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video: How to use the Invitrogen™ QuantiGene™ Plex Data Analysis App on the Thermo Fisher Connect Platform
    Video Player is loading.
    Current Time 0:00
    Duration 12:35
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 12:35
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      QuantiGene Plex Data Analysis Software
      QuantiGene Plex Data Analysis Software is free software available through Connect, the Thermo Fisher Scientific cloud-based platform, that provides easy-to-follow workflows to improve data analysis of QuantiGene Plex assays. Data collected from QuantiGene Plex assays can be exported to Connect and conveniently accessed via web whenever an internet connection is available. Each Connect account offers 1 TB of cloud storage at no charge.
      Access software
      Software capabilities:
      • Control and treatment group assignments
      • Customize QC parameters
      • Analyze data from both 96-well and 384-well plates
      • Perform linearity of dilution calculations
      • Calculation of p-values during fold change analysis
      • Rank normalization genes for your analysis
      • View and export mean and individual sample values
      • Group samples and analyze the fold change in up to five groups
      • Remove samples or targets that do not meet QC criteria
      • Proceed directly to analysis without assigning background wells
      Software workspaceFunctionFeatures
      Setup plate map
      Plate map set up
      Set up plate for analysisAssign:
      • Samples to different groups for comparison
      • Background wells 
      • Wells that should not be included in the analysis
      Normalization
      Normalize data
      Select and edit normalization genes and calculate assay linearity
      • Normalization ranking to calculate the stability measure (M) for each selected gene
      • Calculate assay linearity and % recovery
      Review and edit QC criteria
      Review and edit QC criteria
      View and adjust the thresholds for QC parameters for data analysis
      • Maximum background
      • Limit of Detection
      • Technical Precision (%CV)
      • Saturation
      • Minimum Bead Count
      Run analysis
        Run analysis
      Run and analyze normalized results of differential gene expression analysisCalculate/view:
      • Change in expression of selected targets with respect to the reference sample or reference group and view graph
      Create an account today
      Already have an account? Access the QuantiGene Plex application.
      Learn more about Thermo Fisher Cloud data security
      Software highlights
      Transcriptome Analysis Console
      For enhanced data visualization and analysis beyond simple identification of differential expression, data can be exported from the QuantiGene Plex Data Analysis Software platform into Applied Biosystems Transcriptome Analysis Console (TAC) Software. Find tools to create scatter and volcano plots and perform hierarchical clustering. In addition, link out to publicly available annotations.
      TAC Software capabilities:
      • Perform QC and data normalization
      • Perform statistical tests for differential expression
      • Focus on genes or pathways of interest
      • Explore interactions between coding and non-coding RNA
      • Link out to publicly available annotations
      • Obtain sequence information to design validation experiments
      Download the latest version of TAC Software (64-bit). This version is supported on Microsoft Windows™ 7 (SP 1) and Windows 10 Professional.
      Download  Release notes
      Video: How to set up an Invitrogen™ QuantiGene™ Plex Assay in the Luminex® xPONENT® Software
      Video Player is loading.
      Current Time 0:00
      Duration 8:11
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 8:11
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Video: How to use the Invitrogen™ QuantiGene™ Plex Data Analysis App on the Thermo Fisher Connect Platform
        Video Player is loading.
        Current Time 0:00
        Duration 12:35
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 12:35
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Related products and resources
          For Research Use Only. Not for use in diagnostic procedures.