Protein yield from various cell types using M-PER Mammalian Protein Extraction Reagent
Cells were harvested at 85% confluency, washed twice and collected in ice-cold PBS and counted. For each cell type, 1 x 10^6 cells were pelleted by centrifugation at 2,000 x g for 5 minutes and lysed in 1 mL M-PER Reagent for 5 minutes. The cell lysates were clarified by centrifugation at 14,000 x g for 10 minutes and the supernatant was collected and the protein concentration (µg/million cells) was determined using the Pierce BCA Protein Assay (Part No. 23227).
Cells were  harvested at 85% confluency, washed twice and collected in ice-cold PBS and  counted. For each cell type, 1 x 10^6 cells were pelleted by centrifugation at  2,000 x g for 5 minutes and lysed in 1 mL M-PER Reagent for 5 minutes. The cell lysates were clarified by  centrifugation at 14,000 x g for 10 minutes and the supernatant was collected  and the protein concentration (µg/million cells) was determined using  the Pierce BCA Protein Assay (Part No. 23227).
Related Products
Catalog # Name Size Price (USD) Qty
78501 M-PER™ Mammalian Protein Extraction Reagent, 250 mL Each
368.65
Online Exclusive
408.00
Save 39.35 (10%)
78503 M-PER™ Mammalian Protein Extraction Reagent, 25 mL Each
93.65
Online Exclusive
109.00
Save 15.35 (14%)
78505 M-PER™ Mammalian Protein Extraction Reagent, 1 L Each
966.65
Online Exclusive
1,062.00
Save 95.35 (9%)
Related Images