Lithium-ion battery technology is used in everything from small hand-held electronics to medium sized electric vehicles such as cars, buses and trucks to larger marine vessels and smart grid energy storage systems. Critical to the advancement of the battery is the emergence of higher quality separator film, coating of separator film for higher efficiency, and the critical thickness and weight of the anode and cathode continuous, patch or strip coating.
Thickness measurement and control solutions for lithium-ion batteries help measure the thickness of separator film and the coat weight of both sides of the anode/cathode substrate. For separator film, the latest sensor technology uses full spectrum infrared to measure both the microporous structure and specialty coatings. For the anode and cathode, sensor slot geometry, superior edge to edge measurement and special patch coating measurement algorithms are used to ignore bare substrate, determine key dimensions, and help the industry improve the quality, consistency, and productivity of lithium-ion batteries.