More Filters
View all, or filter by any of the categories below to find resources that are relevant to you
System
Description
SuperScript™ IV UniPrime™ One-Step RT-PCR System
12596100
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 50°C
SuperScript™ IV UniPrime™ One-Step RT-PCR System, colored
12597100
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 50°C
SuperScript™ IV Reverse Transcriptase
18090010
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 50°C
SuperScript™ IV First-Strand Synthesis System
18091050
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 55°C
SuperScript™ IV First-Strand Synthesis System with ezDNase™ Enzyme
18091150
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 55°C
SuperScript™ IV One-Step RT-PCR System
12594025
RNase H activity: No
RT Reaction time: 10 min
Reaction temperature: 55°C
SuperScript™ IV One-Step RT-PCR System with ezDNase™ Enzyme
12595025
RNase H activity: No
RT Reaction time: 10 min
Reaction temperature: 55°C
SuperScript™ IV VILO™ Master Mix
11756500
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 55°C
SuperScript™ IV VILO™ Master Mix with ezDNase™ Enzyme
11766050
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 55°C
SuperScript™ IV CellsDirect™ cDNA Synthesis Kit
11750150
RNase H activity: No
RT Reaction time: 10 mins
Reaction temperature: 50°C
SuperScript™ IV Single Cell/Low Input cDNA PreAmp Kit
11752048
RNase H activity: No
RT Reaction time: 30 mins
Reaction temperature: 50°C
SuperScript™ IV RT-LAMP Master Mix
A51801
RNase H activity: No
RT Reaction time: 10-30 mins
Reaction temperature: 65°C