Comparison of commercially-available phosphatase inhibitors
Kidney extract (25 µL; 0.5 µg/µL) was incubated with a fluorogenic substrate that measures phosphatase activity upon de-phosphorylation. Reactions were incubated for 1 hour at 37ºC and fluorescence was measured. The percent phosphatase inhibition is shown for each phosphatase inhibitor formulation.
Kidney extract (25 µL; 0.5 µg/µL) was incubated with a fluorogenic substrate that measures phosphatase activity upon de-phosphorylation. Reactions were incubated for 1 hour at 37ºC and fluorescence was measured. The percent phosphatase inhibition is shown for each phosphatase inhibitor formulation.
Related Products
Catalog # Name Size Price (USD) Qty
A32957 Pierce™ Phosphatase Inhibitor Mini Tablets, 20 Tablets Each
225.65
Online Exclusive
245.00
Save 19.35 (8%)
Related Images