Search Thermo Fisher Scientific
Live Jurkat cells, or Jurkat cells treated with 0.1% Triton, were mixed and stained with NucBlue® Live and NucGreen® Dead (2 drops per mL) for 15 minutes. Cells were then imaged and analyzed on the EVOS® FL Auto system. NucBlue® Live stains all cells, while NucGreen® Dead stains only dead cells.