Be ready for tomorrow’s most challenging demands
Exceed experimental expectation with the MultiOmics, Structural Biology, and BioPharma Editions of the Thermo Scientific Orbitrap Ascend Editions Tribrid Mass Spectrometer.
With new capabilities for multiplexed quantitative proteomics, native protein characterization, and deep biopharmaceutical analyses, the three new Editions deliver ultimate experimental throughput, versatility, and ease-of-use for your specific needs.
Detect and characterize the most difficult protein complexes and native samples without losing low m/z instrument performance and flexibility with the new Native MS option, including world class Orbitrap detection up to m/z 16,000 and ion isolation in the ion trap and quadrupole up to m/z 8,000, with isolation widths down to m/z 100 and m/z 5, respectively.
Orbitrap Ascend Editions Tribrid MS
Orbitrap Ascend MultiOmics Tribrid MS
Discover more from comprehensive metabolomics to proteomics.
Orbitrap Ascend Structural Biology Tribrid MS
Take advantage of ultimate versatility for native protein characterization.
Orbitrap Ascend BioPharma Tribrid MS
Successfully characterize your most complex biotherapeutics.
For Research Use Only. Not for use in diagnostic procedures.
1x1 image pixel for data collection