Search Thermo Fisher Scientific
Transform your small molecule data, whether a small or large dataset, from liquid chromatography (LC), gas chromatography (GC), ion chromatography (IC), full-scan accurate mass, or MSn data, into actionable insights with Thermo Scientific Compound Discoverer Software. The software offers a fully integrated suite of advanced tools for both known-parent and unknown data processing and interpretation.
This powerful software streamlines compound identification, comparative analyses, and provides extensive filtering and data visualization capabilities. With its easy-to-use workflows, integrated libraries, databases, and robust statistical analysis tools, it enables rapid and precise insights from your valuable data.
Whether your research focuses on metabolomics to stable isotope labeling, environmental and food safety, pharma metabolite or impurity identification, extractables and leachables to forensic or clinical toxicology, Compound Discoverer Software offers an exceptional toolbox to drive precise and efficient data analysis in small-molecule research.
When connected to the Thermo Scientific Ardia Platform, you gain direct access to raw data and result file uploads. Eliminate data management barriers in your laboratory and allows users to collaborate remotely from anywhere.
Reduce the number of mouse clicks | Know your unknowns | Find real differences in your sample sets | Understand biological pathways |
---|---|---|---|
Take control of your data analysis and processing with custom workflows, flexible visualization, and grouping tools. Share results with customizable reporting or transfer your results directly to Thermo Scientific Chromeleon software or Thermo Scientific TraceFinder software for targeted analyses. | Rapidly and confidently identify your unknowns with mass spectral library searching against both the online mzCloud Spectral Library, in-house Thermo Scientific mzVault Spectral Libraries, and numerous built-in annotation tools. | Quickly find significant statistical differences between sample sets. See trends in compounds across a study or identify the key compounds of interest between multiple sample groups using interactively linked displays, including volcano plots, PCA, PLS-DA, and hierarchical clustering. | View pathways using Thermo Scientific Metabolika and BioCyc Databases, and map detected compounds and associated information directly onto pathways. Perform fully untargeted stable isotope labeling experiments and map data onto pathways. |
Discover the power of customizable data analysis workflows with Thermo Scientific Compound Discoverer Software, designed to simplify and enhance your data processing. Tailor each step to suit your specific research needs, ensuring efficient and accurate results.
The right combination of data and software makes all the difference in accurately interpreting complex samples.
Expand the menu to learn about the key features in our Compound Discoverer application.
Studies, whether simple or extensive, produce complex data that contains a wealth of information. To get the most valuable insights from that data, Compound Discoverer Software offers careful data processing, followed by insightful data visualization to speed up reviewing and linking capabilities. With a layered compound tabular view linked directly to a wide array of data visualization tools, you can easily navigate from high level to detailed information on each compound.
Good experimental design is critically important for any analysis, especially for statistical studies to ensure that any potential trends observed occur based upon real changes, rather than those that can be attributed to experimental effects. As such, there are protocols for large-scale studies where the use of pooled quality control (QC) samples are utilized to achieve normalization of these large-scale studies.
Using pooled QC samples, which are analyzed throughout data acquisition, allows for the correction of batch-effects over time. Correction for each compound is performed individually, and multiple methods are available. The displayed correction plot utilizes SERRF QC (Fan et. al). In addition, QC-based correction per compound can be performed based upon a peer-reviewed methodology published by Dunn et. al. in Nature Protocols. Compound Discoverer Software provides the capability to view the impact of any changes to the data pre- and post- correction. In addition, it also supports several general signal normalization tools.
Quality data is the key for discovering the real changes in your data. The software offers a wide range of visualization and data curation tools to assure the quality of your data, and consequently the quality of your conclusions.
Compound Discoverer Software offers a wide range of tools to help with the identification of unknowns. From online and offline spectral libraries, customer user libraries, and compound databases as well as tools for ranking putative identifications like mzLogic and FISh (fragment ion search), the software can help turn more unknowns into knowns.
The software leverages both online and offline library tools for comprehensive data analysis. Online, the mzCloud Spectral Library hosts millions of high-quality curated HRAM MSn spectra on tens of thousands of compounds. Offline libraries, available in the mzVault application format, can assist with PFAS analysis, extractables and leachables, and more. In addition, a compatible version of the NIST Tandem MS library is also available.
The integration of LipidSearch into Compound Discoverer software allows for the detection and annotation of more potential lipids. A database of lipid classes including 96 subclasses along with predicted fragmentation provides a broad range of coverage as well as annotations for the fragmentation spectra of detected lipids. The lipid grade and ID scoring from LipidSearch are included with results giving more information to make decision during data review.
Connectivity between compounds in a complex data set can also be a valuable tool for discovering compounds of interest. Whether by mapping compounds and associated statistical variations against a biological pathway or by connecting related compounds in dynamic molecular networks, Compound Discoverer Software offers a wealth of further identification options.
The fully interactive Molecular Networks visualization browser allows you to view your data in a different way. Identified compounds are shown by nodes (circles) and when a relationship is identified, the nodes are connected. Selecting a node (compound) or connection (transformation) displays pertinent information (right) about the identified compound and the relevant transformation(s). All the visualized data can be interactively filtered using thresholds, data quality information or text search for specific compounds or transformations.
Explore the advanced capabilities and tools within the software that offer researchers enhanced flexibility and precision in their data analysis workflows.
Any software can only do so much, but with the capability to use the Compound Discoverer Scripting node , you can do even more. Use the scripting node in your workflow to automatically export any data from your results, launch your own custom R or Python scripts, and pull the results back into your Compound Discoverer results as a part of the data tables.
In addition to the wide range of data processing tools available to create custom data processing workflows, we also offer a Node Developer Kit (NDK), which allows you to write your own entire custom data processing nodes to include in workflows. The ultimate in flexibility and customizability for those with a talent for software development, or with someone on your team who does.
PyEDS toolPyEDS is a Python library providing a collection of utilities to conveniently access and display results from Thermo Discoverer software including Compound Discoverer software. Using PyEDS, all the hard work of navigating the data hierarchy and recovering the desired information is done automatically, so you can focus more on your research instead of how to read the data.
For the analysis of data acquired using Thermo Scientific GC-Orbitrap-based mass spectrometers, there are two primary workflows, enabled using specific workflow nodes such as Electron Impact (EI) and Chemical Ionization (CI) deconvolution nodes. GC-Orbitrap data can be analyzed using the extensive tools within Compound Discoverer Software to enable confident compound identification, or statistical analysis, for example.
Lee Ferguson's lab developed a PFAS spectral library to enhance rapid identification and quantification of PFAS.
The complete package for small-molecule unknown data processing
We're here to help you maximize your free trial of the Compound Discoverer Software. Contact us if you would like more information on how to use it, or if you would like to have a representative contact you.