Watch this video to learn more about the CryoMed CRF
Video Player is loading.
Current Time 0:00
Duration 4:06
Loaded: 3.99%
Stream Type LIVE
Remaining Time 4:06
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Riptide-Lab
    Integrated OPC UA protocol within the new CryoMed CRF firmware to enable remote unit communication via DeltaV and other systems*.
    • OPC UA = Open Platform Communications Unified Architecture
    • OPC UA: Machine-to-machine communication protocol for industrial automation
    • DeltaV™: Distributed control system for automation systems
    *OPC UA will open-up additional opportunities utilizing in-house automation and IT teams for integration with a data system.
    product features
    CryoMed consistent freezing results
    Enhanced data traceability and customized freezing profiles for consistent temp. performance and real-time monitoring.
    CTS documentation
    Documentation package
    CryoMed CRF with OPC UA comes equipped with a user manual, a certificate of conformance, and a certificate of calibration. 
    Services flyer
    Compliance services
    Service options include installation qualification (IQ), operation qualification (OQ), and temperature mapping.
    Medical Device (available in North America Only)
    • FDA listed Class II medical device, 510K exempt
    • Available in 17 L, 34 L, 48 L
    • With and without built-in thermal printer
    • 100V
    • 120V
    • 220-230V
    General Purpose ( available everywhere)
    • Available in 17 L, 34 L, 48 L
    • With and without built-in thermal printer
    • 100V
    • 120V
    • 220-230V
    Cryomed 3D Tour
    Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›
    CryoMed-blog
    Blog post
    CryoMed Controlled-Rate Freezers that meet 21 CFR Part 11 requirements: Safeguarding sample integrity, traceability and 21 CFR Part 11 compliance.
    Read more >
    Webinar
    On-demand webinar
    A Partnership in Data Traceability: 21 CFR part 11, the CryoMed controlled-rate freezer, and Your Lab/Production Processes
    Watch on-demand >
    Smart Note
    Smart Note
    How can the CryoMed CRF be used to prepare samples and therapies for long-term storage in a cryogenic storage device?
    Read now >
    Style Sheet for Global Design System